Assay Details
Gene Symbol: FRA10AC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_017015662.1 | XP_016871151.1 | 13 - 14 | 1037 | 85 | |
GenBank mRNA | AJ431722.1 | - | 14 - 15 | 1148 | 85 | |
AJ431723.1 | - | 14 - 15 | 1140 | 85 | ||
AJ431724.1 | - | 14 - 15 | 1132 | 85 | ||
AJ431725.1 | - | 13 - 14 | 1068 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1000011454.hg.1 | ENST00000359204.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011454.hg.1 | ENST00000359204.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011454.hg.1 | ENST00000359204.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011454.hg.1 | ENST00000359204.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011454.hg.1 | ENST00000359204.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011454.hg.1 | ENST00000359204.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011454.hg.1 | ENST00000359204.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011454.hg.1 | ENST00000359204.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011454.hg.1 | ENST00000359204.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011454.hg.1 | ENST00000359204.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011454.hg.1 | ENST00000359204.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011454.hg.1 | ENST00000359204.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011454.hg.1 | ENST00000359204.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011454.hg.1 | ENST00000359204.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011454.hg.1 | ENST00000359204.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011454.hg.1 | ENST00000359204.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011454.hg.1 | ENST00000359204.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011454.hg.1 | ENST00000359204.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011454.hg.1 | ENST00000460752.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011454.hg.1 | ENST00000460752.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011454.hg.1 | ENST00000460752.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011454.hg.1 | ENST00000460752.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011454.hg.1 | ENST00000460752.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011454.hg.1 | ENST00000460752.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011454.hg.1 | ENST00000460752.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011454.hg.1 | ENST00000460752.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011454.hg.1 | ENST00000460752.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011454.hg.1 | ENST00000460752.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011454.hg.1 | ENST00000460752.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011454.hg.1 | ENST00000462771.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011454.hg.1 | ENST00000462771.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011454.hg.1 | ENST00000462771.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011454.hg.1 | ENST00000472153.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011454.hg.1 | ENST00000472153.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011454.hg.1 | ENST00000472153.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011454.hg.1 | ENST00000482719.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011454.hg.1 | ENST00000482719.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011454.hg.1 | ENST00000482719.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011454.hg.1 | ENST00000483229.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011454.hg.1 | ENST00000483229.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011454.hg.1 | ENST00000483229.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011454.hg.1 | ENST00000483229.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011454.hg.1 | ENST00000483229.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011454.hg.1 | NM_145246.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000011454.hg.1 | NM_145246.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000011454.hg.1 | NM_145246.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000011454.hg.1 | NM_145246.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000011454.hg.1 | NM_145246.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000011454.hg.1 | NM_145246.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000011454.hg.1 | NM_145246.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000011454.hg.1 | NM_145246.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000011454.hg.1 | NM_145246.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000011454.hg.1 | NM_145246.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000011454.hg.1 | NM_145246.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000011454.hg.1 | NM_145246.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000011454.hg.1 | NM_145246.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000011454.hg.1 | NM_145246.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000011454.hg.1 | NM_145246.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000011454.hg.1 | NM_145246.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000011454.hg.1 | NM_145246.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000011454.hg.1 | NM_145246.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1000011454.hg.1 | ENST00000359204.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011454.hg.1 | ENST00000359204.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011454.hg.1 | ENST00000359204.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011454.hg.1 | ENST00000359204.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011454.hg.1 | ENST00000359204.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011454.hg.1 | ENST00000359204.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011454.hg.1 | ENST00000359204.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011454.hg.1 | ENST00000359204.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011454.hg.1 | ENST00000359204.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011454.hg.1 | ENST00000359204.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011454.hg.1 | ENST00000359204.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011454.hg.1 | ENST00000359204.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011454.hg.1 | ENST00000359204.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011454.hg.1 | ENST00000359204.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011454.hg.1 | ENST00000359204.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011454.hg.1 | ENST00000359204.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011454.hg.1 | ENST00000359204.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011454.hg.1 | ENST00000359204.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011454.hg.1 | ENST00000460752.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011454.hg.1 | ENST00000460752.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011454.hg.1 | ENST00000460752.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011454.hg.1 | ENST00000460752.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011454.hg.1 | ENST00000460752.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011454.hg.1 | ENST00000460752.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011454.hg.1 | ENST00000460752.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011454.hg.1 | ENST00000460752.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011454.hg.1 | ENST00000460752.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011454.hg.1 | ENST00000460752.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011454.hg.1 | ENST00000460752.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011454.hg.1 | ENST00000462771.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011454.hg.1 | ENST00000462771.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011454.hg.1 | ENST00000462771.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011454.hg.1 | ENST00000472153.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011454.hg.1 | ENST00000472153.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011454.hg.1 | ENST00000472153.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011454.hg.1 | ENST00000482719.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011454.hg.1 | ENST00000482719.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011454.hg.1 | ENST00000482719.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011454.hg.1 | ENST00000483229.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011454.hg.1 | ENST00000483229.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011454.hg.1 | ENST00000483229.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011454.hg.1 | ENST00000483229.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011454.hg.1 | ENST00000483229.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011454.hg.1 | NM_145246.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1000011454.hg.1 | NM_145246.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1000011454.hg.1 | NM_145246.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1000011454.hg.1 | NM_145246.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1000011454.hg.1 | NM_145246.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1000011454.hg.1 | NM_145246.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1000011454.hg.1 | NM_145246.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1000011454.hg.1 | NM_145246.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1000011454.hg.1 | NM_145246.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1000011454.hg.1 | NM_145246.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1000011454.hg.1 | NM_145246.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1000011454.hg.1 | NM_145246.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1000011454.hg.1 | NM_145246.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1000011454.hg.1 | NM_145246.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1000011454.hg.1 | NM_145246.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1000011454.hg.1 | NM_145246.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1000011454.hg.1 | NM_145246.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1000011454.hg.1 | NM_145246.4 | Refseq |
