Assay Details
Gene Symbol: WASF3-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AW070512.1 | - | 1 - 2 | 266 | 112 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1300008414.hg.1 | ENST00000413063.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008414.hg.1 | ENST00000413063.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008414.hg.1 | ENST00000413063.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008414.hg.1 | ENST00000413063.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008414.hg.1 | ENST00000413063.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008414.hg.1 | ENST00000413063.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008414.hg.1 | ENST00000413063.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008414.hg.1 | ENST00000585599.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008414.hg.1 | ENST00000585599.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008414.hg.1 | ENST00000585599.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008414.hg.1 | ENST00000585599.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008414.hg.1 | ENST00000585599.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008414.hg.1 | ENST00000586418.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008414.hg.1 | ENST00000586418.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008414.hg.1 | ENST00000586418.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008414.hg.1 | ENST00000586418.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008414.hg.1 | ENST00000586418.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008414.hg.1 | ENST00000586418.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008414.hg.1 | ENST00000625894.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008414.hg.1 | ENST00000625894.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008414.hg.1 | ENST00000626038.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008414.hg.1 | ENST00000626038.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008414.hg.1 | ENST00000626779.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008414.hg.1 | ENST00000626779.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008414.hg.1 | ENST00000626779.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008414.hg.1 | ENST00000626779.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008414.hg.1 | ENST00000627315.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008414.hg.1 | ENST00000627315.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008414.hg.1 | ENST00000627315.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008414.hg.1 | ENST00000627315.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008414.hg.1 | ENST00000630958.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008414.hg.1 | ENST00000630958.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008414.hg.1 | ENST00000630958.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008414.hg.1 | ENST00000630958.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008414.hg.1 | ENST00000630958.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008414.hg.1 | ENST00000413063.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008414.hg.1 | ENST00000413063.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008414.hg.1 | ENST00000413063.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008414.hg.1 | ENST00000413063.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008414.hg.1 | ENST00000413063.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008414.hg.1 | ENST00000413063.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008414.hg.1 | ENST00000413063.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008414.hg.1 | ENST00000585599.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008414.hg.1 | ENST00000585599.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008414.hg.1 | ENST00000585599.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008414.hg.1 | ENST00000585599.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008414.hg.1 | ENST00000585599.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008414.hg.1 | ENST00000586418.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008414.hg.1 | ENST00000586418.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008414.hg.1 | ENST00000586418.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008414.hg.1 | ENST00000586418.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008414.hg.1 | ENST00000586418.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008414.hg.1 | ENST00000586418.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008414.hg.1 | ENST00000625894.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008414.hg.1 | ENST00000625894.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008414.hg.1 | ENST00000626038.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008414.hg.1 | ENST00000626038.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008414.hg.1 | ENST00000626779.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008414.hg.1 | ENST00000626779.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008414.hg.1 | ENST00000626779.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008414.hg.1 | ENST00000626779.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008414.hg.1 | ENST00000627315.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008414.hg.1 | ENST00000627315.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008414.hg.1 | ENST00000627315.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008414.hg.1 | ENST00000627315.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008414.hg.1 | ENST00000630958.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008414.hg.1 | ENST00000630958.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008414.hg.1 | ENST00000630958.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008414.hg.1 | ENST00000630958.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008414.hg.1 | ENST00000630958.2 | Ensembl |
