Assay Details
| Gene Symbol: LOC102724474 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_011537532.2 | XP_011535834.1 | 2 - 3 | 695 | 62 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC102724474| Gene Symbol: LOC102724474 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_011537532.2 | XP_011535834.1 | 2 - 3 | 695 | 62 | |
Set Membership: |
|