Assay Details
| Gene Symbol: HOMEZ | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_020834.2 | NP_065885.2 | 2 | 1915 | 126 | |
| GenBank mRNA | AB037864.1 | - | 2 | 1838 | 126 | |
| AF463523.1 | - | 2 | 1836 | 126 | ||
| BC130392.1 | - | 1 | 1737 | 40146423 | 126 | |
More Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1400010716.hg.1 | ENST00000357460.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010716.hg.1 | ENST00000357460.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010716.hg.1 | ENST00000357460.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010716.hg.1 | ENST00000357460.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010716.hg.1 | ENST00000357460.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010716.hg.1 | ENST00000357460.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010716.hg.1 | ENST00000357460.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010716.hg.1 | ENST00000357460.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010716.hg.1 | ENST00000357460.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010716.hg.1 | ENST00000357460.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010716.hg.1 | ENST00000357460.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010716.hg.1 | ENST00000558278.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010716.hg.1 | ENST00000558278.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010716.hg.1 | ENST00000558278.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010716.hg.1 | ENST00000558278.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010716.hg.1 | ENST00000558278.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010716.hg.1 | ENST00000606731.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010716.hg.1 | ENST00000606731.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010716.hg.1 | NM_020834.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010716.hg.1 | NM_020834.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010716.hg.1 | NM_020834.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010716.hg.1 | NM_020834.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010716.hg.1 | NM_020834.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010716.hg.1 | NM_020834.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010716.hg.1 | NM_020834.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010716.hg.1 | NM_020834.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010716.hg.1 | NM_020834.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400010716.hg.1 | NM_020834.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011457.hg.1 | ENST00000565635.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011457.hg.1 | ENST00000565635.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011457.hg.1 | ENST00000566864.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011457.hg.1 | ENST00000566864.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011457.hg.1 | ENST00000566864.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011457.hg.1 | ENST00000568739.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011457.hg.1 | ENST00000568739.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011457.hg.1 | ENST00000568739.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011457.hg.1 | ENST00000568739.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011457.hg.1 | ENST00000568739.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011457.hg.1 | ENST00000568739.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011457.hg.1 | ENST00000568739.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011457.hg.1 | ENST00000568739.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011457.hg.1 | ENST00000568739.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011457.hg.1 | ENST00000568739.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011457.hg.1 | ENST00000568739.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011457.hg.1 | ENST00000568739.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011457.hg.1 | ENST00000568739.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011457.hg.1 | ENST00000568739.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011457.hg.1 | ENST00000569162.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011457.hg.1 | ENST00000569162.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011457.hg.1 | ENST00000569162.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011457.hg.1 | ENST00000569162.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011457.hg.1 | ENST00000569162.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011457.hg.1 | ENST00000569162.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600011457.hg.1 | ENST00000569162.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010716.hg.1 | ENST00000357460.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010716.hg.1 | ENST00000357460.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010716.hg.1 | ENST00000357460.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010716.hg.1 | ENST00000357460.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010716.hg.1 | ENST00000357460.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010716.hg.1 | ENST00000357460.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010716.hg.1 | ENST00000357460.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010716.hg.1 | ENST00000357460.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010716.hg.1 | ENST00000357460.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010716.hg.1 | ENST00000357460.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010716.hg.1 | ENST00000357460.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010716.hg.1 | ENST00000558278.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010716.hg.1 | ENST00000558278.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010716.hg.1 | ENST00000558278.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010716.hg.1 | ENST00000558278.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010716.hg.1 | ENST00000558278.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010716.hg.1 | ENST00000606731.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010716.hg.1 | ENST00000606731.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400010716.hg.1 | NM_020834.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010716.hg.1 | NM_020834.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010716.hg.1 | NM_020834.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010716.hg.1 | NM_020834.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010716.hg.1 | NM_020834.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010716.hg.1 | NM_020834.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010716.hg.1 | NM_020834.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010716.hg.1 | NM_020834.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010716.hg.1 | NM_020834.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400010716.hg.1 | NM_020834.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600011457.hg.1 | ENST00000565635.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011457.hg.1 | ENST00000565635.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011457.hg.1 | ENST00000566864.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011457.hg.1 | ENST00000566864.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011457.hg.1 | ENST00000566864.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011457.hg.1 | ENST00000568739.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011457.hg.1 | ENST00000568739.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011457.hg.1 | ENST00000568739.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011457.hg.1 | ENST00000568739.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011457.hg.1 | ENST00000568739.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011457.hg.1 | ENST00000568739.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011457.hg.1 | ENST00000568739.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011457.hg.1 | ENST00000568739.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011457.hg.1 | ENST00000568739.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011457.hg.1 | ENST00000568739.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011457.hg.1 | ENST00000568739.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011457.hg.1 | ENST00000568739.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011457.hg.1 | ENST00000568739.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011457.hg.1 | ENST00000568739.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011457.hg.1 | ENST00000569162.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011457.hg.1 | ENST00000569162.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011457.hg.1 | ENST00000569162.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011457.hg.1 | ENST00000569162.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011457.hg.1 | ENST00000569162.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011457.hg.1 | ENST00000569162.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600011457.hg.1 | ENST00000569162.1 | Ensembl |

HOMEZ
Genomic Map