Assay Details
Gene Symbol: PNP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_000270.3 | NP_000261.2 | 6 | 1425 | 127 | |
GenBank mRNA | AK098544.1 | - | 6 | 1406 | 127 | |
AK126154.1 | - | 2 | 2497 | 127 | ||
BC106074.1 | - | 6 | 1421 | 3859664 | 127 | |
EU794649.1 | - | 6 | 1421 | 127 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
nucleobase-containing compound metabolic process
inosine catabolic process
purine nucleotide catabolic process
nicotinamide riboside catabolic process
immune response
NAD biosynthesis via nicotinamide riboside salvage pathway
urate biosynthetic process
positive regulation of T cell proliferation
response to drug
purine-containing compound salvage
positive regulation of alpha-beta T cell differentiation
interleukin-2 secretion
nucleoside binding
purine nucleobase binding
purine-nucleoside phosphorylase activity
drug binding
phosphate ion binding
nucleobase-containing compound metabolic process
inosine catabolic process
purine nucleotide catabolic process
nicotinamide riboside catabolic process
immune response
NAD biosynthesis via nicotinamide riboside salvage pathway
urate biosynthetic process
positive regulation of T cell proliferation
response to drug
purine-containing compound salvage
positive regulation of alpha-beta T cell differentiation
interleukin-2 secretion
nucleoside binding
purine nucleobase binding
purine-nucleoside phosphorylase activity
drug binding
phosphate ion binding
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1400008592.hg.1 | ENST00000554678.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400008592.hg.1 | ENST00000554678.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400006521.hg.1 | ENST00000361505.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400006521.hg.1 | ENST00000361505.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400006521.hg.1 | ENST00000361505.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400006521.hg.1 | ENST00000361505.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400006521.hg.1 | ENST00000361505.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400006521.hg.1 | ENST00000361505.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400006521.hg.1 | ENST00000361505.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400006521.hg.1 | ENST00000361505.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400006521.hg.1 | ENST00000361505.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400006521.hg.1 | ENST00000361505.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400006521.hg.1 | ENST00000361505.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400006521.hg.1 | ENST00000361505.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400006521.hg.1 | ENST00000553418.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400006521.hg.1 | ENST00000553418.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400006521.hg.1 | ENST00000553418.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400006521.hg.1 | ENST00000553418.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400006521.hg.1 | ENST00000553418.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400006521.hg.1 | ENST00000553591.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400006521.hg.1 | ENST00000553591.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400006521.hg.1 | ENST00000553591.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400006521.hg.1 | ENST00000553591.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400006521.hg.1 | ENST00000553591.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400006521.hg.1 | ENST00000553591.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400006521.hg.1 | ENST00000554056.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400006521.hg.1 | ENST00000554056.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400006521.hg.1 | ENST00000554056.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400006521.hg.1 | ENST00000554056.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400006521.hg.1 | ENST00000554056.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400006521.hg.1 | ENST00000554056.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400006521.hg.1 | ENST00000554056.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400006521.hg.1 | ENST00000554056.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400006521.hg.1 | ENST00000554056.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400006521.hg.1 | ENST00000554056.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400006521.hg.1 | ENST00000554056.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400006521.hg.1 | ENST00000554056.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400006521.hg.1 | ENST00000554065.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400006521.hg.1 | ENST00000554065.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400006521.hg.1 | ENST00000554065.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400006521.hg.1 | ENST00000554065.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400006521.hg.1 | ENST00000556293.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400006521.hg.1 | ENST00000556293.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400006521.hg.1 | ENST00000556293.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400006521.hg.1 | ENST00000556754.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400006521.hg.1 | ENST00000556754.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400006521.hg.1 | ENST00000556754.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400006521.hg.1 | ENST00000556754.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400006521.hg.1 | ENST00000556754.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400006521.hg.1 | ENST00000556754.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400006521.hg.1 | ENST00000556754.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400006521.hg.1 | ENST00000556754.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400006521.hg.1 | ENST00000556754.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400006521.hg.1 | ENST00000556754.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400006521.hg.1 | ENST00000556754.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400006521.hg.1 | ENST00000556754.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400006521.hg.1 | ENST00000556754.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400006521.hg.1 | ENST00000557229.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400006521.hg.1 | ENST00000557229.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400006521.hg.1 | ENST00000557229.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400006521.hg.1 | ENST00000557229.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400006521.hg.1 | ENST00000557229.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400006521.hg.1 | ENST00000557229.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400006521.hg.1 | ENST00000557229.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400006521.hg.1 | ENST00000557229.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400006521.hg.1 | NM_000270.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400006521.hg.1 | NM_000270.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400006521.hg.1 | NM_000270.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400006521.hg.1 | NM_000270.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400006521.hg.1 | NM_000270.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400006521.hg.1 | NM_000270.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400006521.hg.1 | NM_000270.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400006521.hg.1 | NM_000270.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400006521.hg.1 | NM_000270.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400006521.hg.1 | NM_000270.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400006521.hg.1 | NM_000270.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400006521.hg.1 | NM_000270.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400006521.hg.1 | NM_000270.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1400006521.hg.1 | ENST00000361505.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400006521.hg.1 | ENST00000361505.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400006521.hg.1 | ENST00000361505.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400006521.hg.1 | ENST00000361505.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400006521.hg.1 | ENST00000361505.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400006521.hg.1 | ENST00000361505.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400006521.hg.1 | ENST00000361505.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400006521.hg.1 | ENST00000361505.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400006521.hg.1 | ENST00000361505.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400006521.hg.1 | ENST00000361505.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400006521.hg.1 | ENST00000361505.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400006521.hg.1 | ENST00000361505.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400006521.hg.1 | ENST00000553418.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400006521.hg.1 | ENST00000553418.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400006521.hg.1 | ENST00000553418.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400006521.hg.1 | ENST00000553418.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400006521.hg.1 | ENST00000553418.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400006521.hg.1 | ENST00000553591.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400006521.hg.1 | ENST00000553591.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400006521.hg.1 | ENST00000553591.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400006521.hg.1 | ENST00000553591.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400006521.hg.1 | ENST00000553591.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400006521.hg.1 | ENST00000553591.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400006521.hg.1 | ENST00000554056.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400006521.hg.1 | ENST00000554056.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400006521.hg.1 | ENST00000554056.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400006521.hg.1 | ENST00000554056.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400006521.hg.1 | ENST00000554056.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400006521.hg.1 | ENST00000554056.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400006521.hg.1 | ENST00000554056.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400006521.hg.1 | ENST00000554056.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400006521.hg.1 | ENST00000554056.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400006521.hg.1 | ENST00000554056.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400006521.hg.1 | ENST00000554056.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400006521.hg.1 | ENST00000554056.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400006521.hg.1 | ENST00000554065.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400006521.hg.1 | ENST00000554065.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400006521.hg.1 | ENST00000554065.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400006521.hg.1 | ENST00000554065.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400006521.hg.1 | ENST00000556293.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400006521.hg.1 | ENST00000556293.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400006521.hg.1 | ENST00000556293.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400006521.hg.1 | ENST00000556754.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400006521.hg.1 | ENST00000556754.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400006521.hg.1 | ENST00000556754.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400006521.hg.1 | ENST00000556754.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400006521.hg.1 | ENST00000556754.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400006521.hg.1 | ENST00000556754.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400006521.hg.1 | ENST00000556754.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400006521.hg.1 | ENST00000556754.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400006521.hg.1 | ENST00000556754.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400006521.hg.1 | ENST00000556754.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400006521.hg.1 | ENST00000556754.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400006521.hg.1 | ENST00000556754.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400006521.hg.1 | ENST00000556754.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400006521.hg.1 | ENST00000557229.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400006521.hg.1 | ENST00000557229.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400006521.hg.1 | ENST00000557229.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400006521.hg.1 | ENST00000557229.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400006521.hg.1 | ENST00000557229.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400006521.hg.1 | ENST00000557229.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400006521.hg.1 | ENST00000557229.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400006521.hg.1 | ENST00000557229.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400006521.hg.1 | NM_000270.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1400006521.hg.1 | NM_000270.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1400006521.hg.1 | NM_000270.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1400006521.hg.1 | NM_000270.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1400006521.hg.1 | NM_000270.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1400006521.hg.1 | NM_000270.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1400006521.hg.1 | NM_000270.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1400006521.hg.1 | NM_000270.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1400006521.hg.1 | NM_000270.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1400006521.hg.1 | NM_000270.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1400006521.hg.1 | NM_000270.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1400006521.hg.1 | NM_000270.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1400006521.hg.1 | NM_000270.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1400008592.hg.1 | ENST00000554678.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400008592.hg.1 | ENST00000554678.1 | Ensembl |
