Assay Details
| Gene Symbol: LOC283588 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK055958.1 | - | 1 | 1618 | 85 | |
| AK095276.1 | - | 1 | 2367 | 85 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC283588| Gene Symbol: LOC283588 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK055958.1 | - | 1 | 1618 | 85 | |
| AK095276.1 | - | 1 | 2367 | 85 | ||
Set Membership: |
|