Assay Details
Gene Symbol: MAPK6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005254537.2 | XP_005254594.1 | 1 - 2 | 102 | 84 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005254537.2 transcripts.
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1500010743.hg.1 | ENST00000261845.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010743.hg.1 | ENST00000261845.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010743.hg.1 | ENST00000261845.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010743.hg.1 | ENST00000261845.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010743.hg.1 | ENST00000261845.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010743.hg.1 | ENST00000261845.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010743.hg.1 | ENST00000261845.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010743.hg.1 | ENST00000261845.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010743.hg.1 | ENST00000261845.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010743.hg.1 | ENST00000261845.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010743.hg.1 | ENST00000261845.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010743.hg.1 | ENST00000261845.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010743.hg.1 | ENST00000261845.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010743.hg.1 | ENST00000261845.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010743.hg.1 | ENST00000558100.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010743.hg.1 | ENST00000558100.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010743.hg.1 | ENST00000558100.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010743.hg.1 | ENST00000558100.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010743.hg.1 | ENST00000558841.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010743.hg.1 | ENST00000558841.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010743.hg.1 | ENST00000558841.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010743.hg.1 | ENST00000558841.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010743.hg.1 | ENST00000558841.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010743.hg.1 | ENST00000558891.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010743.hg.1 | ENST00000558891.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010743.hg.1 | ENST00000558891.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010743.hg.1 | ENST00000558891.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010743.hg.1 | ENST00000560774.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010743.hg.1 | ENST00000560774.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010743.hg.1 | ENST00000560774.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010743.hg.1 | NM_002748.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010743.hg.1 | NM_002748.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010743.hg.1 | NM_002748.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010743.hg.1 | NM_002748.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010743.hg.1 | NM_002748.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010743.hg.1 | NM_002748.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010743.hg.1 | NM_002748.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010743.hg.1 | NM_002748.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010743.hg.1 | NM_002748.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010743.hg.1 | NM_002748.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010743.hg.1 | NM_002748.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010743.hg.1 | NM_002748.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010743.hg.1 | NM_002748.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1500010743.hg.1 | ENST00000261845.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010743.hg.1 | ENST00000261845.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010743.hg.1 | ENST00000261845.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010743.hg.1 | ENST00000261845.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010743.hg.1 | ENST00000261845.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010743.hg.1 | ENST00000261845.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010743.hg.1 | ENST00000261845.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010743.hg.1 | ENST00000261845.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010743.hg.1 | ENST00000261845.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010743.hg.1 | ENST00000261845.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010743.hg.1 | ENST00000261845.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010743.hg.1 | ENST00000261845.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010743.hg.1 | ENST00000261845.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010743.hg.1 | ENST00000261845.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010743.hg.1 | ENST00000558100.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010743.hg.1 | ENST00000558100.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010743.hg.1 | ENST00000558100.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010743.hg.1 | ENST00000558100.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010743.hg.1 | ENST00000558841.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010743.hg.1 | ENST00000558841.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010743.hg.1 | ENST00000558841.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010743.hg.1 | ENST00000558841.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010743.hg.1 | ENST00000558841.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010743.hg.1 | ENST00000558891.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010743.hg.1 | ENST00000558891.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010743.hg.1 | ENST00000558891.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010743.hg.1 | ENST00000558891.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010743.hg.1 | ENST00000560774.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010743.hg.1 | ENST00000560774.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010743.hg.1 | ENST00000560774.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010743.hg.1 | NM_002748.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1500010743.hg.1 | NM_002748.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1500010743.hg.1 | NM_002748.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1500010743.hg.1 | NM_002748.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1500010743.hg.1 | NM_002748.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1500010743.hg.1 | NM_002748.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1500010743.hg.1 | NM_002748.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1500010743.hg.1 | NM_002748.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1500010743.hg.1 | NM_002748.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1500010743.hg.1 | NM_002748.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1500010743.hg.1 | NM_002748.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1500010743.hg.1 | NM_002748.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1500010743.hg.1 | NM_002748.3 | Refseq |
