Assay Details
| Gene Symbol: LINC01579 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | DB014757.1 | - | 1 - 2 | 191 | 69 | |
More Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1500010932.hg.1 | ENST00000553818.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010932.hg.1 | ENST00000553818.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010932.hg.1 | ENST00000553818.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010932.hg.1 | ENST00000553818.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010932.hg.1 | ENST00000554261.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010932.hg.1 | ENST00000554261.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010932.hg.1 | ENST00000554261.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010932.hg.1 | ENST00000555023.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010932.hg.1 | ENST00000555023.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010932.hg.1 | ENST00000555023.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010932.hg.1 | ENST00000557481.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010932.hg.1 | ENST00000557481.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010932.hg.1 | ENST00000557481.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010932.hg.1 | ENST00000557481.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010932.hg.1 | ENST00000557481.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010932.hg.1 | ENST00000557481.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010932.hg.1 | ENST00000557481.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010932.hg.1 | ENST00000557481.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010932.hg.1 | ENST00000557481.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010932.hg.1 | ENST00000557481.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010932.hg.1 | ENST00000557481.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010932.hg.1 | ENST00000557481.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010932.hg.1 | ENST00000557481.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010932.hg.1 | ENST00000557481.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010932.hg.1 | ENST00000557715.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010932.hg.1 | ENST00000557715.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010932.hg.1 | ENST00000557715.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010932.hg.1 | ENST00000557777.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010932.hg.1 | ENST00000557777.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010932.hg.1 | ENST00000557777.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010932.hg.1 | ENST00000557777.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010932.hg.1 | ENST00000557777.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010932.hg.1 | ENST00000557777.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010932.hg.1 | ENST00000553818.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010932.hg.1 | ENST00000553818.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010932.hg.1 | ENST00000553818.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010932.hg.1 | ENST00000553818.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010932.hg.1 | ENST00000554261.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010932.hg.1 | ENST00000554261.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010932.hg.1 | ENST00000554261.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010932.hg.1 | ENST00000555023.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010932.hg.1 | ENST00000555023.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010932.hg.1 | ENST00000555023.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010932.hg.1 | ENST00000557481.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010932.hg.1 | ENST00000557481.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010932.hg.1 | ENST00000557481.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010932.hg.1 | ENST00000557481.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010932.hg.1 | ENST00000557481.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010932.hg.1 | ENST00000557481.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010932.hg.1 | ENST00000557481.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010932.hg.1 | ENST00000557481.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010932.hg.1 | ENST00000557481.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010932.hg.1 | ENST00000557481.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010932.hg.1 | ENST00000557481.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010932.hg.1 | ENST00000557481.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010932.hg.1 | ENST00000557481.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010932.hg.1 | ENST00000557481.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010932.hg.1 | ENST00000557715.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010932.hg.1 | ENST00000557715.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010932.hg.1 | ENST00000557715.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010932.hg.1 | ENST00000557777.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010932.hg.1 | ENST00000557777.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010932.hg.1 | ENST00000557777.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010932.hg.1 | ENST00000557777.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010932.hg.1 | ENST00000557777.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010932.hg.1 | ENST00000557777.1 | Ensembl |

LINC01579
Genomic Map