Assay Details
| Gene Symbol: NBEAP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AL832227.1 | - | 4 - 5 | 809 | 128 | |
More Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1500008627.hg.1 | ENST00000554452.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008627.hg.1 | ENST00000554452.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008627.hg.1 | ENST00000554452.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008627.hg.1 | ENST00000554452.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008627.hg.1 | ENST00000554452.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008627.hg.1 | ENST00000554452.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008627.hg.1 | ENST00000554452.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008627.hg.1 | ENST00000554452.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008627.hg.1 | ENST00000554452.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008627.hg.1 | ENST00000556948.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008627.hg.1 | ENST00000556948.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008627.hg.1 | ENST00000556948.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008627.hg.1 | ENST00000556948.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008627.hg.1 | ENST00000564214.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008627.hg.1 | ENST00000564214.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008627.hg.1 | ENST00000564214.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008627.hg.1 | NR_027992.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008627.hg.1 | NR_027992.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008627.hg.1 | NR_027992.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010068.hg.1 | ENST00000327925.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010068.hg.1 | ENST00000327925.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010068.hg.1 | ENST00000327925.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010068.hg.1 | ENST00000327925.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010068.hg.1 | ENST00000327925.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010068.hg.1 | ENST00000327925.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010068.hg.1 | ENST00000327925.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010068.hg.1 | ENST00000327925.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010068.hg.1 | ENST00000539177.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010068.hg.1 | ENST00000539177.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010068.hg.1 | ENST00000539177.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010068.hg.1 | ENST00000539484.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010068.hg.1 | ENST00000539484.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010068.hg.1 | ENST00000539484.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010068.hg.1 | ENST00000539484.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010068.hg.1 | ENST00000539484.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010068.hg.1 | ENST00000539484.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010068.hg.1 | NM_001004306.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010068.hg.1 | NM_001004306.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010068.hg.1 | NM_001004306.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010068.hg.1 | NM_001004306.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010068.hg.1 | NM_001004306.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010068.hg.1 | NM_001004306.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010068.hg.1 | NM_001004306.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010068.hg.1 | NM_001004306.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500008627.hg.1 | ENST00000554452.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008627.hg.1 | ENST00000554452.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008627.hg.1 | ENST00000554452.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008627.hg.1 | ENST00000554452.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008627.hg.1 | ENST00000554452.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008627.hg.1 | ENST00000554452.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008627.hg.1 | ENST00000554452.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008627.hg.1 | ENST00000554452.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008627.hg.1 | ENST00000554452.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008627.hg.1 | ENST00000556948.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008627.hg.1 | ENST00000556948.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008627.hg.1 | ENST00000556948.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008627.hg.1 | ENST00000556948.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008627.hg.1 | ENST00000564214.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008627.hg.1 | ENST00000564214.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008627.hg.1 | ENST00000564214.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008627.hg.1 | NR_027992.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500008627.hg.1 | NR_027992.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500008627.hg.1 | NR_027992.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010068.hg.1 | ENST00000327925.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010068.hg.1 | ENST00000327925.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010068.hg.1 | ENST00000327925.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010068.hg.1 | ENST00000327925.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010068.hg.1 | ENST00000327925.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010068.hg.1 | ENST00000327925.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010068.hg.1 | ENST00000327925.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010068.hg.1 | ENST00000327925.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010068.hg.1 | ENST00000539177.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010068.hg.1 | ENST00000539177.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010068.hg.1 | ENST00000539177.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010068.hg.1 | ENST00000539484.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010068.hg.1 | ENST00000539484.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010068.hg.1 | ENST00000539484.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010068.hg.1 | ENST00000539484.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010068.hg.1 | ENST00000539484.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010068.hg.1 | ENST00000539484.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010068.hg.1 | NM_001004306.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010068.hg.1 | NM_001004306.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010068.hg.1 | NM_001004306.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010068.hg.1 | NM_001004306.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010068.hg.1 | NM_001004306.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010068.hg.1 | NM_001004306.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010068.hg.1 | NM_001004306.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010068.hg.1 | NM_001004306.2 | Refseq |

NBEAP1
Genomic Map