Assay Details
Gene Symbol: PATL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_011521340.2 | XP_011519642.1 | 1 - 2 | 371 | 102 | |
GenBank mRNA | AK130783.1 | - | 1 - 2 | 298 | 102 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_011521340.2 transcripts.
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1500009263.hg.1 | ENST00000434130.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009263.hg.1 | ENST00000434130.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009263.hg.1 | ENST00000434130.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009263.hg.1 | ENST00000434130.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009263.hg.1 | ENST00000434130.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009263.hg.1 | ENST00000434130.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009263.hg.1 | ENST00000434130.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009263.hg.1 | ENST00000434130.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009263.hg.1 | ENST00000434130.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009263.hg.1 | ENST00000434130.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009263.hg.1 | ENST00000434130.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009263.hg.1 | ENST00000434130.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009263.hg.1 | ENST00000434130.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009263.hg.1 | ENST00000434130.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009263.hg.1 | ENST00000434130.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009263.hg.1 | ENST00000434130.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009263.hg.1 | ENST00000434130.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009263.hg.1 | ENST00000434130.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009263.hg.1 | ENST00000558159.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009263.hg.1 | ENST00000558159.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009263.hg.1 | ENST00000558159.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009263.hg.1 | ENST00000558481.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009263.hg.1 | ENST00000558481.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009263.hg.1 | ENST00000558481.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009263.hg.1 | ENST00000558481.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009263.hg.1 | ENST00000558481.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009263.hg.1 | ENST00000558481.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009263.hg.1 | ENST00000558481.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009263.hg.1 | ENST00000558481.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009263.hg.1 | ENST00000558481.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009263.hg.1 | ENST00000558573.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009263.hg.1 | ENST00000558573.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009263.hg.1 | ENST00000558573.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009263.hg.1 | ENST00000558573.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009263.hg.1 | ENST00000558573.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009263.hg.1 | ENST00000558573.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009263.hg.1 | ENST00000558809.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009263.hg.1 | ENST00000558809.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009263.hg.1 | ENST00000558809.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009263.hg.1 | ENST00000560110.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009263.hg.1 | ENST00000560110.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009263.hg.1 | ENST00000560110.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009263.hg.1 | ENST00000560110.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009263.hg.1 | ENST00000560110.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009263.hg.1 | ENST00000560110.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009263.hg.1 | ENST00000560110.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009263.hg.1 | ENST00000560775.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009263.hg.1 | ENST00000560775.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009263.hg.1 | ENST00000560775.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009263.hg.1 | ENST00000560775.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009263.hg.1 | ENST00000560775.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009263.hg.1 | ENST00000560775.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009263.hg.1 | ENST00000560775.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009263.hg.1 | ENST00000560775.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009263.hg.1 | ENST00000560775.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009263.hg.1 | ENST00000560775.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009263.hg.1 | ENST00000560775.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009263.hg.1 | ENST00000560775.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009263.hg.1 | ENST00000560775.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009263.hg.1 | ENST00000560775.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009263.hg.1 | ENST00000560775.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009263.hg.1 | ENST00000560775.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009263.hg.1 | ENST00000560775.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009263.hg.1 | ENST00000560775.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009263.hg.1 | ENST00000560780.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009263.hg.1 | ENST00000560780.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009263.hg.1 | ENST00000560780.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009263.hg.1 | ENST00000560780.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009263.hg.1 | ENST00000560780.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009263.hg.1 | ENST00000560780.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009263.hg.1 | ENST00000560780.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009263.hg.1 | ENST00000560780.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009263.hg.1 | ENST00000560780.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009263.hg.1 | ENST00000560780.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009263.hg.1 | ENST00000560780.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009263.hg.1 | ENST00000560780.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009263.hg.1 | ENST00000560780.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009263.hg.1 | ENST00000560780.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009263.hg.1 | ENST00000560780.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009263.hg.1 | ENST00000560780.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009263.hg.1 | ENST00000560780.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009263.hg.1 | ENST00000561082.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009263.hg.1 | ENST00000561082.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009263.hg.1 | ENST00000561082.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009263.hg.1 | ENST00000561082.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009263.hg.1 | NM_001145112.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500009263.hg.1 | NM_001145112.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500009263.hg.1 | NM_001145112.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500009263.hg.1 | NM_001145112.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500009263.hg.1 | NM_001145112.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500009263.hg.1 | NM_001145112.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500009263.hg.1 | NM_001145112.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500009263.hg.1 | NM_001145112.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500009263.hg.1 | NM_001145112.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500009263.hg.1 | NM_001145112.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500009263.hg.1 | NM_001145112.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500009263.hg.1 | NM_001145112.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500009263.hg.1 | NM_001145112.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500009263.hg.1 | NM_001145112.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500009263.hg.1 | NM_001145112.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500009263.hg.1 | NM_001145112.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500009263.hg.1 | NM_001145112.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500009263.hg.1 | NM_001145112.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500009263.hg.1 | ENST00000434130.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009263.hg.1 | ENST00000434130.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009263.hg.1 | ENST00000434130.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009263.hg.1 | ENST00000434130.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009263.hg.1 | ENST00000434130.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009263.hg.1 | ENST00000434130.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009263.hg.1 | ENST00000434130.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009263.hg.1 | ENST00000434130.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009263.hg.1 | ENST00000434130.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009263.hg.1 | ENST00000434130.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009263.hg.1 | ENST00000434130.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009263.hg.1 | ENST00000434130.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009263.hg.1 | ENST00000434130.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009263.hg.1 | ENST00000434130.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009263.hg.1 | ENST00000434130.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009263.hg.1 | ENST00000434130.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009263.hg.1 | ENST00000434130.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009263.hg.1 | ENST00000434130.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009263.hg.1 | ENST00000558159.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009263.hg.1 | ENST00000558159.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009263.hg.1 | ENST00000558159.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009263.hg.1 | ENST00000558481.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009263.hg.1 | ENST00000558481.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009263.hg.1 | ENST00000558481.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009263.hg.1 | ENST00000558481.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009263.hg.1 | ENST00000558481.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009263.hg.1 | ENST00000558481.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009263.hg.1 | ENST00000558481.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009263.hg.1 | ENST00000558481.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009263.hg.1 | ENST00000558481.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009263.hg.1 | ENST00000558573.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009263.hg.1 | ENST00000558573.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009263.hg.1 | ENST00000558573.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009263.hg.1 | ENST00000558573.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009263.hg.1 | ENST00000558573.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009263.hg.1 | ENST00000558573.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009263.hg.1 | ENST00000558809.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009263.hg.1 | ENST00000558809.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009263.hg.1 | ENST00000558809.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009263.hg.1 | ENST00000560110.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009263.hg.1 | ENST00000560110.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009263.hg.1 | ENST00000560110.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009263.hg.1 | ENST00000560110.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009263.hg.1 | ENST00000560110.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009263.hg.1 | ENST00000560110.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009263.hg.1 | ENST00000560110.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009263.hg.1 | ENST00000560775.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009263.hg.1 | ENST00000560775.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009263.hg.1 | ENST00000560775.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009263.hg.1 | ENST00000560775.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009263.hg.1 | ENST00000560775.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009263.hg.1 | ENST00000560775.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009263.hg.1 | ENST00000560775.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009263.hg.1 | ENST00000560775.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009263.hg.1 | ENST00000560775.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009263.hg.1 | ENST00000560775.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009263.hg.1 | ENST00000560775.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009263.hg.1 | ENST00000560775.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009263.hg.1 | ENST00000560775.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009263.hg.1 | ENST00000560775.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009263.hg.1 | ENST00000560775.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009263.hg.1 | ENST00000560775.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009263.hg.1 | ENST00000560775.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009263.hg.1 | ENST00000560775.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009263.hg.1 | ENST00000560780.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009263.hg.1 | ENST00000560780.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009263.hg.1 | ENST00000560780.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009263.hg.1 | ENST00000560780.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009263.hg.1 | ENST00000560780.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009263.hg.1 | ENST00000560780.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009263.hg.1 | ENST00000560780.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009263.hg.1 | ENST00000560780.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009263.hg.1 | ENST00000560780.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009263.hg.1 | ENST00000560780.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009263.hg.1 | ENST00000560780.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009263.hg.1 | ENST00000560780.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009263.hg.1 | ENST00000560780.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009263.hg.1 | ENST00000560780.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009263.hg.1 | ENST00000560780.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009263.hg.1 | ENST00000560780.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009263.hg.1 | ENST00000560780.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009263.hg.1 | ENST00000561082.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009263.hg.1 | ENST00000561082.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009263.hg.1 | ENST00000561082.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009263.hg.1 | ENST00000561082.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009263.hg.1 | NM_001145112.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500009263.hg.1 | NM_001145112.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500009263.hg.1 | NM_001145112.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500009263.hg.1 | NM_001145112.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500009263.hg.1 | NM_001145112.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500009263.hg.1 | NM_001145112.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500009263.hg.1 | NM_001145112.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500009263.hg.1 | NM_001145112.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500009263.hg.1 | NM_001145112.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500009263.hg.1 | NM_001145112.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500009263.hg.1 | NM_001145112.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500009263.hg.1 | NM_001145112.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500009263.hg.1 | NM_001145112.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500009263.hg.1 | NM_001145112.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500009263.hg.1 | NM_001145112.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500009263.hg.1 | NM_001145112.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500009263.hg.1 | NM_001145112.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500009263.hg.1 | NM_001145112.1 | Refseq |
