Assay Details
Gene Symbol: GPR176 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001271854.1 | NP_001258783.1 | 1 - 2 | 1038 | 103 | |
GenBank mRNA | BC067106.1 | - | 1 - 2 | 371 | 6645890 | 103 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1500010854.hg.1 | ENST00000299092.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010854.hg.1 | ENST00000299092.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010854.hg.1 | ENST00000299092.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010854.hg.1 | ENST00000299092.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010854.hg.1 | ENST00000299092.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010854.hg.1 | ENST00000299092.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010854.hg.1 | ENST00000299092.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010854.hg.1 | ENST00000299092.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010854.hg.1 | ENST00000299092.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010854.hg.1 | ENST00000299092.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010854.hg.1 | ENST00000543580.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010854.hg.1 | ENST00000543580.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010854.hg.1 | ENST00000543580.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010854.hg.1 | ENST00000543580.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010854.hg.1 | ENST00000543580.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010854.hg.1 | ENST00000543580.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010854.hg.1 | ENST00000543580.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010854.hg.1 | ENST00000543580.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010854.hg.1 | ENST00000543580.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010854.hg.1 | ENST00000558041.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010854.hg.1 | ENST00000558041.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010854.hg.1 | ENST00000558041.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010854.hg.1 | ENST00000558041.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010854.hg.1 | ENST00000560729.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010854.hg.1 | ENST00000560729.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010854.hg.1 | ENST00000560729.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010854.hg.1 | ENST00000560729.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010854.hg.1 | ENST00000560729.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010854.hg.1 | ENST00000560729.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010854.hg.1 | ENST00000560729.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010854.hg.1 | ENST00000561100.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010854.hg.1 | ENST00000561100.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010854.hg.1 | ENST00000561100.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010854.hg.1 | ENST00000561100.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010854.hg.1 | ENST00000561100.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010854.hg.1 | ENST00000561100.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010854.hg.1 | ENST00000561100.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010854.hg.1 | ENST00000561100.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010854.hg.1 | ENST00000561100.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010854.hg.1 | ENST00000561100.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010854.hg.1 | NM_001271854.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010854.hg.1 | NM_001271854.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010854.hg.1 | NM_001271854.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010854.hg.1 | NM_001271854.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010854.hg.1 | NM_001271854.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010854.hg.1 | NM_001271854.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010854.hg.1 | NM_001271854.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010854.hg.1 | NM_001271854.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010854.hg.1 | NM_001271854.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010854.hg.1 | NM_001271854.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010854.hg.1 | NM_001271854.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010854.hg.1 | NM_001271855.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010854.hg.1 | NM_001271855.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010854.hg.1 | NM_001271855.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010854.hg.1 | NM_001271855.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010854.hg.1 | NM_001271855.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010854.hg.1 | NM_001271855.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010854.hg.1 | NM_001271855.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010854.hg.1 | NM_001271855.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010854.hg.1 | NM_001271855.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010854.hg.1 | NM_007223.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010854.hg.1 | NM_007223.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010854.hg.1 | NM_007223.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010854.hg.1 | NM_007223.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010854.hg.1 | NM_007223.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010854.hg.1 | NM_007223.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010854.hg.1 | NM_007223.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010854.hg.1 | NM_007223.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010854.hg.1 | NM_007223.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010854.hg.1 | NM_007223.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010854.hg.1 | NM_007223.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1500010854.hg.1 | ENST00000299092.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010854.hg.1 | ENST00000299092.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010854.hg.1 | ENST00000299092.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010854.hg.1 | ENST00000299092.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010854.hg.1 | ENST00000299092.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010854.hg.1 | ENST00000299092.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010854.hg.1 | ENST00000299092.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010854.hg.1 | ENST00000299092.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010854.hg.1 | ENST00000299092.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010854.hg.1 | ENST00000299092.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010854.hg.1 | ENST00000543580.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010854.hg.1 | ENST00000543580.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010854.hg.1 | ENST00000543580.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010854.hg.1 | ENST00000543580.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010854.hg.1 | ENST00000543580.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010854.hg.1 | ENST00000543580.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010854.hg.1 | ENST00000543580.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010854.hg.1 | ENST00000543580.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010854.hg.1 | ENST00000543580.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010854.hg.1 | ENST00000558041.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010854.hg.1 | ENST00000558041.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010854.hg.1 | ENST00000558041.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010854.hg.1 | ENST00000558041.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010854.hg.1 | ENST00000560729.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010854.hg.1 | ENST00000560729.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010854.hg.1 | ENST00000560729.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010854.hg.1 | ENST00000560729.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010854.hg.1 | ENST00000560729.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010854.hg.1 | ENST00000560729.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010854.hg.1 | ENST00000560729.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010854.hg.1 | ENST00000561100.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010854.hg.1 | ENST00000561100.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010854.hg.1 | ENST00000561100.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010854.hg.1 | ENST00000561100.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010854.hg.1 | ENST00000561100.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010854.hg.1 | ENST00000561100.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010854.hg.1 | ENST00000561100.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010854.hg.1 | ENST00000561100.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010854.hg.1 | ENST00000561100.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010854.hg.1 | ENST00000561100.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010854.hg.1 | NM_001271854.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500010854.hg.1 | NM_001271854.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500010854.hg.1 | NM_001271854.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500010854.hg.1 | NM_001271854.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500010854.hg.1 | NM_001271854.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500010854.hg.1 | NM_001271854.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500010854.hg.1 | NM_001271854.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500010854.hg.1 | NM_001271854.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500010854.hg.1 | NM_001271854.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500010854.hg.1 | NM_001271854.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500010854.hg.1 | NM_001271854.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500010854.hg.1 | NM_001271855.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500010854.hg.1 | NM_001271855.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500010854.hg.1 | NM_001271855.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500010854.hg.1 | NM_001271855.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500010854.hg.1 | NM_001271855.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500010854.hg.1 | NM_001271855.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500010854.hg.1 | NM_001271855.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500010854.hg.1 | NM_001271855.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500010854.hg.1 | NM_001271855.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500010854.hg.1 | NM_007223.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1500010854.hg.1 | NM_007223.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1500010854.hg.1 | NM_007223.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1500010854.hg.1 | NM_007223.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1500010854.hg.1 | NM_007223.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1500010854.hg.1 | NM_007223.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1500010854.hg.1 | NM_007223.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1500010854.hg.1 | NM_007223.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1500010854.hg.1 | NM_007223.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1500010854.hg.1 | NM_007223.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1500010854.hg.1 | NM_007223.2 | Refseq |
