Assay Details
Gene Symbol: LOC102723344 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_120375.1 | 5 - 6 | 395 | 99 | ||
GenBank mRNA | BG675369.1 | - | 4 - 5 | 395 | 100 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1500007524.hg.1 | ENST00000559379.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007524.hg.1 | ENST00000559379.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007524.hg.1 | ENST00000559379.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007524.hg.1 | ENST00000559379.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007524.hg.1 | ENST00000559379.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007524.hg.1 | ENST00000559379.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007524.hg.1 | ENST00000559379.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007524.hg.1 | ENST00000559379.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007524.hg.1 | ENST00000559379.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007524.hg.1 | ENST00000559379.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007524.hg.1 | ENST00000559379.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007524.hg.1 | ENST00000559379.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007524.hg.1 | ENST00000559379.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007524.hg.1 | ENST00000559379.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007524.hg.1 | ENST00000559379.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007524.hg.1 | ENST00000559379.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007524.hg.1 | ENST00000559379.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007524.hg.1 | ENST00000559379.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007524.hg.1 | ENST00000559821.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007524.hg.1 | ENST00000559821.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007524.hg.1 | ENST00000559821.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007524.hg.1 | ENST00000559821.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007524.hg.1 | ENST00000559821.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007524.hg.1 | ENST00000559821.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007524.hg.1 | ENST00000559821.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007524.hg.1 | ENST00000559821.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007524.hg.1 | ENST00000559821.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007524.hg.1 | ENST00000559821.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007524.hg.1 | ENST00000559821.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007524.hg.1 | ENST00000559821.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007524.hg.1 | ENST00000559821.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007524.hg.1 | ENST00000559821.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007524.hg.1 | ENST00000559821.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007524.hg.1 | ENST00000559821.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007524.hg.1 | ENST00000559821.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007524.hg.1 | ENST00000559821.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007524.hg.1 | NR_120375.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500007524.hg.1 | NR_120375.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500007524.hg.1 | NR_120375.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500007524.hg.1 | NR_120375.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500007524.hg.1 | NR_120375.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500007524.hg.1 | NR_120375.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500007524.hg.1 | NR_120375.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500007524.hg.1 | NR_120375.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500007524.hg.1 | NR_120375.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500007524.hg.1 | ENST00000559379.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007524.hg.1 | ENST00000559379.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007524.hg.1 | ENST00000559379.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007524.hg.1 | ENST00000559379.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007524.hg.1 | ENST00000559379.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007524.hg.1 | ENST00000559379.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007524.hg.1 | ENST00000559379.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007524.hg.1 | ENST00000559379.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007524.hg.1 | ENST00000559379.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007524.hg.1 | ENST00000559379.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007524.hg.1 | ENST00000559379.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007524.hg.1 | ENST00000559379.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007524.hg.1 | ENST00000559379.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007524.hg.1 | ENST00000559379.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007524.hg.1 | ENST00000559379.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007524.hg.1 | ENST00000559379.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007524.hg.1 | ENST00000559379.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007524.hg.1 | ENST00000559379.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007524.hg.1 | ENST00000559821.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007524.hg.1 | ENST00000559821.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007524.hg.1 | ENST00000559821.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007524.hg.1 | ENST00000559821.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007524.hg.1 | ENST00000559821.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007524.hg.1 | ENST00000559821.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007524.hg.1 | ENST00000559821.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007524.hg.1 | ENST00000559821.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007524.hg.1 | ENST00000559821.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007524.hg.1 | ENST00000559821.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007524.hg.1 | ENST00000559821.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007524.hg.1 | ENST00000559821.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007524.hg.1 | ENST00000559821.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007524.hg.1 | ENST00000559821.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007524.hg.1 | ENST00000559821.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007524.hg.1 | ENST00000559821.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007524.hg.1 | ENST00000559821.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007524.hg.1 | ENST00000559821.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007524.hg.1 | NR_120375.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500007524.hg.1 | NR_120375.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500007524.hg.1 | NR_120375.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500007524.hg.1 | NR_120375.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500007524.hg.1 | NR_120375.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500007524.hg.1 | NR_120375.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500007524.hg.1 | NR_120375.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500007524.hg.1 | NR_120375.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500007524.hg.1 | NR_120375.1 | Refseq |
