Assay Details
Gene Symbol: RAP1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001010935.2 | NP_001010935.1 | 9 | 3054 | 109 | |
NM_001291896.1 | NP_001278825.1 | 8 | 3009 | 109 | ||
NM_002884.3 | NP_002875.1 | 8 | 2960 | 109 | ||
GenBank mRNA | AK094737.1 | - | 1 | 420 | 109 | |
BU189979.1 | - | 1 | 98 | 109 | ||
BX473371.1 | - | 1 | 516 | 109 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
activation of MAPKK activity
signal transduction
positive regulation of neuron projection development
protein transport
microvillus assembly
Rap protein signal transduction
negative regulation of collagen biosynthetic process
cellular response to drug
nerve growth factor signaling pathway
positive regulation of GTPase activity
positive regulation of protein kinase activity
positive regulation of glucose import
positive regulation of phagocytosis
regulation of insulin secretion
establishment of endothelial barrier
positive regulation of ERK1 and ERK2 cascade
cellular response to cAMP
cellular response to glucose stimulus
response to antineoplastic agent
liver regeneration
regulation of cell junction assembly
cellular response to nerve growth factor stimulus
negative regulation of synaptic vesicle exocytosis
positive regulation of vasculogenesis
GTPase activity
protein binding
GTP binding
protein transporter activity
Ras GTPase binding
Rap guanyl-nucleotide exchange factor activity
protein complex binding
activation of MAPKK activity
signal transduction
positive regulation of neuron projection development
protein transport
microvillus assembly
Rap protein signal transduction
negative regulation of collagen biosynthetic process
cellular response to drug
nerve growth factor signaling pathway
positive regulation of GTPase activity
positive regulation of protein kinase activity
positive regulation of glucose import
positive regulation of phagocytosis
regulation of insulin secretion
establishment of endothelial barrier
positive regulation of ERK1 and ERK2 cascade
cellular response to cAMP
cellular response to glucose stimulus
response to antineoplastic agent
liver regeneration
regulation of cell junction assembly
cellular response to nerve growth factor stimulus
negative regulation of synaptic vesicle exocytosis
positive regulation of vasculogenesis
GTPase activity
protein binding
GTP binding
protein transporter activity
Ras GTPase binding
Rap guanyl-nucleotide exchange factor activity
protein complex binding
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0100009418.hg.1 | ENST00000356415.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009418.hg.1 | ENST00000356415.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009418.hg.1 | ENST00000356415.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009418.hg.1 | ENST00000356415.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009418.hg.1 | ENST00000356415.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009418.hg.1 | ENST00000356415.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009418.hg.1 | ENST00000356415.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009418.hg.1 | ENST00000356415.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009418.hg.1 | ENST00000356415.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009418.hg.1 | ENST00000356415.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009418.hg.1 | ENST00000356415.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009418.hg.1 | ENST00000356415.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009418.hg.1 | ENST00000369709.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009418.hg.1 | ENST00000369709.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009418.hg.1 | ENST00000369709.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009418.hg.1 | ENST00000369709.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009418.hg.1 | ENST00000369709.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009418.hg.1 | ENST00000369709.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009418.hg.1 | ENST00000369709.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009418.hg.1 | ENST00000369709.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009418.hg.1 | ENST00000369709.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009418.hg.1 | ENST00000369709.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009418.hg.1 | ENST00000369709.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009418.hg.1 | ENST00000369709.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009418.hg.1 | ENST00000369709.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009418.hg.1 | ENST00000369709.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009418.hg.1 | ENST00000369709.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009418.hg.1 | ENST00000433097.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009418.hg.1 | ENST00000433097.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009418.hg.1 | ENST00000433097.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009418.hg.1 | ENST00000433097.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009418.hg.1 | ENST00000433097.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009418.hg.1 | ENST00000433097.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009418.hg.1 | ENST00000433097.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009418.hg.1 | ENST00000433097.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009418.hg.1 | ENST00000494982.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009418.hg.1 | ENST00000494982.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009418.hg.1 | ENST00000494982.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009418.hg.1 | ENST00000494982.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009418.hg.1 | ENST00000494982.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009418.hg.1 | ENST00000494982.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009418.hg.1 | ENST00000494982.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009418.hg.1 | ENST00000494982.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009418.hg.1 | ENST00000494982.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009418.hg.1 | ENST00000494982.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009418.hg.1 | ENST00000494982.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009418.hg.1 | ENST00000494982.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009418.hg.1 | NM_001010935.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009418.hg.1 | NM_001010935.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009418.hg.1 | NM_001010935.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009418.hg.1 | NM_001010935.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009418.hg.1 | NM_001010935.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009418.hg.1 | NM_001010935.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009418.hg.1 | NM_001010935.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009418.hg.1 | NM_001010935.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009418.hg.1 | NM_001010935.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009418.hg.1 | NM_001010935.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009418.hg.1 | NM_001010935.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009418.hg.1 | NM_001010935.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009418.hg.1 | NM_001010935.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009418.hg.1 | NM_001010935.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009418.hg.1 | NM_001010935.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009418.hg.1 | NM_001010935.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009418.hg.1 | NM_001291896.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009418.hg.1 | NM_001291896.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009418.hg.1 | NM_001291896.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009418.hg.1 | NM_001291896.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009418.hg.1 | NM_001291896.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009418.hg.1 | NM_001291896.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009418.hg.1 | NM_001291896.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009418.hg.1 | NM_001291896.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009418.hg.1 | NM_001291896.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009418.hg.1 | NM_001291896.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009418.hg.1 | NM_001291896.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009418.hg.1 | NM_001291896.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009418.hg.1 | NM_001291896.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009418.hg.1 | NM_001291896.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009418.hg.1 | NM_001291896.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009418.hg.1 | NM_002884.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009418.hg.1 | NM_002884.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009418.hg.1 | NM_002884.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009418.hg.1 | NM_002884.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009418.hg.1 | NM_002884.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009418.hg.1 | NM_002884.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009418.hg.1 | NM_002884.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009418.hg.1 | NM_002884.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009418.hg.1 | NM_002884.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009418.hg.1 | NM_002884.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009418.hg.1 | NM_002884.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009418.hg.1 | NM_002884.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009418.hg.1 | NM_002884.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009418.hg.1 | NM_002884.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009418.hg.1 | NM_002884.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009418.hg.1 | ENST00000356415.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009418.hg.1 | ENST00000356415.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009418.hg.1 | ENST00000356415.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009418.hg.1 | ENST00000356415.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009418.hg.1 | ENST00000356415.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009418.hg.1 | ENST00000356415.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009418.hg.1 | ENST00000356415.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009418.hg.1 | ENST00000356415.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009418.hg.1 | ENST00000356415.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009418.hg.1 | ENST00000356415.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009418.hg.1 | ENST00000356415.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009418.hg.1 | ENST00000356415.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009418.hg.1 | ENST00000369709.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009418.hg.1 | ENST00000369709.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009418.hg.1 | ENST00000369709.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009418.hg.1 | ENST00000369709.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009418.hg.1 | ENST00000369709.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009418.hg.1 | ENST00000369709.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009418.hg.1 | ENST00000369709.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009418.hg.1 | ENST00000369709.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009418.hg.1 | ENST00000369709.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009418.hg.1 | ENST00000369709.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009418.hg.1 | ENST00000369709.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009418.hg.1 | ENST00000369709.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009418.hg.1 | ENST00000369709.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009418.hg.1 | ENST00000369709.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009418.hg.1 | ENST00000369709.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009418.hg.1 | ENST00000433097.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009418.hg.1 | ENST00000433097.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009418.hg.1 | ENST00000433097.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009418.hg.1 | ENST00000433097.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009418.hg.1 | ENST00000433097.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009418.hg.1 | ENST00000433097.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009418.hg.1 | ENST00000433097.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009418.hg.1 | ENST00000433097.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009418.hg.1 | ENST00000494982.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009418.hg.1 | ENST00000494982.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009418.hg.1 | ENST00000494982.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009418.hg.1 | ENST00000494982.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009418.hg.1 | ENST00000494982.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009418.hg.1 | ENST00000494982.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009418.hg.1 | ENST00000494982.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009418.hg.1 | ENST00000494982.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009418.hg.1 | ENST00000494982.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009418.hg.1 | ENST00000494982.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009418.hg.1 | ENST00000494982.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009418.hg.1 | ENST00000494982.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009418.hg.1 | NM_001010935.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009418.hg.1 | NM_001010935.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009418.hg.1 | NM_001010935.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009418.hg.1 | NM_001010935.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009418.hg.1 | NM_001010935.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009418.hg.1 | NM_001010935.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009418.hg.1 | NM_001010935.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009418.hg.1 | NM_001010935.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009418.hg.1 | NM_001010935.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009418.hg.1 | NM_001010935.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009418.hg.1 | NM_001010935.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009418.hg.1 | NM_001010935.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009418.hg.1 | NM_001010935.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009418.hg.1 | NM_001010935.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009418.hg.1 | NM_001010935.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009418.hg.1 | NM_001010935.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009418.hg.1 | NM_001291896.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009418.hg.1 | NM_001291896.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009418.hg.1 | NM_001291896.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009418.hg.1 | NM_001291896.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009418.hg.1 | NM_001291896.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009418.hg.1 | NM_001291896.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009418.hg.1 | NM_001291896.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009418.hg.1 | NM_001291896.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009418.hg.1 | NM_001291896.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009418.hg.1 | NM_001291896.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009418.hg.1 | NM_001291896.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009418.hg.1 | NM_001291896.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009418.hg.1 | NM_001291896.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009418.hg.1 | NM_001291896.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009418.hg.1 | NM_001291896.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009418.hg.1 | NM_002884.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009418.hg.1 | NM_002884.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009418.hg.1 | NM_002884.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009418.hg.1 | NM_002884.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009418.hg.1 | NM_002884.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009418.hg.1 | NM_002884.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009418.hg.1 | NM_002884.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009418.hg.1 | NM_002884.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009418.hg.1 | NM_002884.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009418.hg.1 | NM_002884.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009418.hg.1 | NM_002884.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009418.hg.1 | NM_002884.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009418.hg.1 | NM_002884.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009418.hg.1 | NM_002884.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009418.hg.1 | NM_002884.3 | Refseq |
