Assay Details
| Gene Symbol: BIRC6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016252.3 | NP_057336.3 | 18 - 19 | 4247 | 91 | |
| XM_005264449.4 | XP_005264506.2 | 18 - 19 | 4410 | 91 | ||
| XM_005264450.4 | XP_005264507.2 | 18 - 19 | 4410 | 91 | ||
| XM_005264451.4 | XP_005264508.2 | 18 - 19 | 4410 | 91 | ||
| XM_005264452.4 | XP_005264509.2 | 18 - 19 | 4113 | 91 | ||
| XM_005264453.4 | XP_005264510.2 | 18 - 19 | 4113 | 91 | ||
| XM_005264454.4 | XP_005264511.2 | 18 - 19 | 4410 | 91 | ||
| XM_005264455.4 | XP_005264512.2 | 17 - 18 | 4278 | 91 | ||
| XM_006712054.3 | XP_006712117.1 | 18 - 19 | 4410 | 91 | ||
| XM_006712055.3 | XP_006712118.1 | 18 - 19 | 4410 | 91 | ||
| XM_006712056.3 | XP_006712119.1 | 18 - 19 | 4410 | 91 | ||
| XM_011533003.2 | XP_011531305.1 | 18 - 19 | 3855 | 91 | ||
| XM_011533005.2 | XP_011531307.1 | 18 - 19 | 4410 | 91 | ||
| XM_017004556.1 | XP_016860045.1 | 18 - 19 | 4410 | 91 | ||
| XM_017004557.1 | XP_016860046.1 | 18 - 19 | 4410 | 91 | ||
| XM_017004558.1 | XP_016860047.1 | 18 - 19 | 4368 | 91 | ||
| XM_017004559.1 | XP_016860048.1 | 18 - 19 | 4410 | 91 | ||
| XM_017004560.1 | XP_016860049.1 | 18 - 19 | 4410 | 91 | ||
| GenBank mRNA | AF265555.1 | - | 18 - 19 | 4029 | 91 | |
More Information
Set Membership: |
|

BIRC6
Genomic Map