Assay Details
Gene Symbol: ODF3B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BC127934.1 | - | 1 | 753 | 60 | |
S72487.1 | - | 1 | 457 | 60 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC2200009363.hg.1 | ENST00000329363.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009363.hg.1 | ENST00000329363.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009363.hg.1 | ENST00000329363.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009363.hg.1 | ENST00000329363.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009363.hg.1 | ENST00000329363.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009363.hg.1 | ENST00000329363.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009363.hg.1 | ENST00000329363.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009363.hg.1 | ENST00000329363.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009363.hg.1 | ENST00000329363.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009363.hg.1 | ENST00000329363.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009363.hg.1 | ENST00000329363.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009363.hg.1 | ENST00000401779.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009363.hg.1 | ENST00000401779.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009363.hg.1 | ENST00000401779.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009363.hg.1 | ENST00000401779.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009363.hg.1 | ENST00000401779.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009363.hg.1 | ENST00000401779.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009363.hg.1 | ENST00000401779.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009363.hg.1 | ENST00000401779.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009363.hg.1 | ENST00000401779.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009363.hg.1 | ENST00000401779.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009363.hg.1 | ENST00000403326.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009363.hg.1 | ENST00000403326.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009363.hg.1 | ENST00000403326.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009363.hg.1 | ENST00000403326.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009363.hg.1 | ENST00000403326.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009363.hg.1 | ENST00000403326.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009363.hg.1 | ENST00000403326.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009363.hg.1 | ENST00000405135.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009363.hg.1 | ENST00000405135.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009363.hg.1 | ENST00000405135.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009363.hg.1 | ENST00000405135.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009363.hg.1 | ENST00000405135.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009363.hg.1 | ENST00000405135.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009363.hg.1 | ENST00000405135.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009363.hg.1 | ENST00000405135.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009363.hg.1 | ENST00000405135.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009363.hg.1 | ENST00000405135.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009363.hg.1 | ENST00000405135.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009363.hg.1 | ENST00000405135.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009363.hg.1 | ENST00000428989.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009363.hg.1 | ENST00000428989.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009363.hg.1 | ENST00000428989.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009363.hg.1 | ENST00000428989.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009363.hg.1 | ENST00000428989.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009363.hg.1 | ENST00000428989.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009363.hg.1 | ENST00000428989.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009363.hg.1 | ENST00000428989.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009363.hg.1 | ENST00000437588.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009363.hg.1 | ENST00000437588.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009363.hg.1 | ENST00000437588.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009363.hg.1 | ENST00000437588.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009363.hg.1 | ENST00000463472.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009363.hg.1 | ENST00000463472.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009363.hg.1 | ENST00000463472.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009363.hg.1 | ENST00000463472.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009363.hg.1 | ENST00000463472.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009363.hg.1 | ENST00000463472.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009363.hg.1 | ENST00000463472.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009363.hg.1 | ENST00000463472.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009363.hg.1 | ENST00000469660.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009363.hg.1 | ENST00000469660.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009363.hg.1 | ENST00000469660.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009363.hg.1 | ENST00000469660.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009363.hg.1 | ENST00000469660.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009363.hg.1 | ENST00000469660.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200009363.hg.1 | NM_001014440.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200009363.hg.1 | NM_001014440.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200009363.hg.1 | NM_001014440.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200009363.hg.1 | NM_001014440.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200009363.hg.1 | NM_001014440.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200009363.hg.1 | NM_001014440.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200009363.hg.1 | NM_001014440.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200009363.hg.1 | NM_001014440.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200009363.hg.1 | NM_001014440.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200009363.hg.1 | NM_001014440.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200009363.hg.1 | NM_001014440.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2200009363.hg.1 | ENST00000329363.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009363.hg.1 | ENST00000329363.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009363.hg.1 | ENST00000329363.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009363.hg.1 | ENST00000329363.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009363.hg.1 | ENST00000329363.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009363.hg.1 | ENST00000329363.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009363.hg.1 | ENST00000329363.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009363.hg.1 | ENST00000329363.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009363.hg.1 | ENST00000329363.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009363.hg.1 | ENST00000329363.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009363.hg.1 | ENST00000329363.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009363.hg.1 | ENST00000401779.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009363.hg.1 | ENST00000401779.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009363.hg.1 | ENST00000401779.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009363.hg.1 | ENST00000401779.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009363.hg.1 | ENST00000401779.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009363.hg.1 | ENST00000401779.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009363.hg.1 | ENST00000401779.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009363.hg.1 | ENST00000401779.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009363.hg.1 | ENST00000401779.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009363.hg.1 | ENST00000401779.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009363.hg.1 | ENST00000403326.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009363.hg.1 | ENST00000403326.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009363.hg.1 | ENST00000403326.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009363.hg.1 | ENST00000403326.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009363.hg.1 | ENST00000403326.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009363.hg.1 | ENST00000403326.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009363.hg.1 | ENST00000403326.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009363.hg.1 | ENST00000405135.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009363.hg.1 | ENST00000405135.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009363.hg.1 | ENST00000405135.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009363.hg.1 | ENST00000405135.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009363.hg.1 | ENST00000405135.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009363.hg.1 | ENST00000405135.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009363.hg.1 | ENST00000405135.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009363.hg.1 | ENST00000405135.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009363.hg.1 | ENST00000405135.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009363.hg.1 | ENST00000405135.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009363.hg.1 | ENST00000405135.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009363.hg.1 | ENST00000405135.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009363.hg.1 | ENST00000428989.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009363.hg.1 | ENST00000428989.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009363.hg.1 | ENST00000428989.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009363.hg.1 | ENST00000428989.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009363.hg.1 | ENST00000428989.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009363.hg.1 | ENST00000428989.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009363.hg.1 | ENST00000428989.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009363.hg.1 | ENST00000428989.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009363.hg.1 | ENST00000437588.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009363.hg.1 | ENST00000437588.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009363.hg.1 | ENST00000437588.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009363.hg.1 | ENST00000437588.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009363.hg.1 | ENST00000463472.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009363.hg.1 | ENST00000463472.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009363.hg.1 | ENST00000463472.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009363.hg.1 | ENST00000463472.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009363.hg.1 | ENST00000463472.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009363.hg.1 | ENST00000463472.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009363.hg.1 | ENST00000463472.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009363.hg.1 | ENST00000463472.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009363.hg.1 | ENST00000469660.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009363.hg.1 | ENST00000469660.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009363.hg.1 | ENST00000469660.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009363.hg.1 | ENST00000469660.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009363.hg.1 | ENST00000469660.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009363.hg.1 | ENST00000469660.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200009363.hg.1 | NM_001014440.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2200009363.hg.1 | NM_001014440.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2200009363.hg.1 | NM_001014440.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2200009363.hg.1 | NM_001014440.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2200009363.hg.1 | NM_001014440.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2200009363.hg.1 | NM_001014440.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2200009363.hg.1 | NM_001014440.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2200009363.hg.1 | NM_001014440.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2200009363.hg.1 | NM_001014440.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2200009363.hg.1 | NM_001014440.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2200009363.hg.1 | NM_001014440.3 | Refseq |
