Assay Details
Gene Symbol: ADORA2A-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_028484.2 | 7 | 2681 | 100 | ||
GenBank mRNA | AK091970.1 | - | 3 | 1844 | 100 | |
BF509218.1 | - | 1 | 100 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
