Assay Details
Gene Symbol: EPM2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_011536116.1 | XP_011534418.1 | 1 - 2 | 32 | 82 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_011536116.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0600013458.hg.1 | ENST00000367519.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013458.hg.1 | ENST00000367519.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013458.hg.1 | ENST00000367519.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013458.hg.1 | ENST00000367519.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013458.hg.1 | ENST00000367519.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013458.hg.1 | ENST00000367519.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013458.hg.1 | ENST00000367519.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013458.hg.1 | ENST00000367519.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013458.hg.1 | ENST00000367519.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013458.hg.1 | ENST00000367519.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013458.hg.1 | ENST00000367519.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013458.hg.1 | ENST00000367519.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013458.hg.1 | ENST00000367519.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013458.hg.1 | ENST00000435470.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013458.hg.1 | ENST00000435470.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013458.hg.1 | ENST00000435470.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013458.hg.1 | ENST00000435470.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013458.hg.1 | ENST00000435470.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013458.hg.1 | ENST00000435470.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013458.hg.1 | ENST00000435470.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013458.hg.1 | ENST00000450221.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013458.hg.1 | ENST00000450221.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013458.hg.1 | ENST00000450221.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013458.hg.1 | ENST00000450221.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013458.hg.1 | ENST00000450221.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013458.hg.1 | ENST00000450221.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013458.hg.1 | ENST00000461700.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013458.hg.1 | ENST00000461700.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013458.hg.1 | ENST00000461700.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013458.hg.1 | ENST00000461700.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013458.hg.1 | ENST00000461700.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013458.hg.1 | ENST00000461700.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013458.hg.1 | ENST00000461700.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013458.hg.1 | ENST00000461700.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013458.hg.1 | ENST00000489412.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013458.hg.1 | ENST00000489412.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013458.hg.1 | ENST00000489412.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013458.hg.1 | ENST00000489412.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013458.hg.1 | ENST00000496228.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013458.hg.1 | ENST00000496228.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013458.hg.1 | ENST00000496228.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013458.hg.1 | ENST00000496228.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013458.hg.1 | ENST00000496228.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013458.hg.1 | ENST00000496228.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013458.hg.1 | ENST00000496228.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013458.hg.1 | ENST00000496228.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013458.hg.1 | ENST00000496228.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013458.hg.1 | ENST00000496228.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013458.hg.1 | ENST00000611340.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013458.hg.1 | ENST00000611340.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013458.hg.1 | ENST00000611340.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013458.hg.1 | ENST00000611340.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013458.hg.1 | ENST00000611340.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013458.hg.1 | ENST00000611340.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013458.hg.1 | ENST00000611340.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013458.hg.1 | ENST00000611340.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013458.hg.1 | ENST00000611340.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013458.hg.1 | ENST00000611340.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013458.hg.1 | ENST00000618445.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013458.hg.1 | ENST00000618445.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013458.hg.1 | ENST00000618445.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013458.hg.1 | ENST00000618445.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013458.hg.1 | ENST00000618445.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013458.hg.1 | ENST00000618445.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013458.hg.1 | ENST00000618445.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013458.hg.1 | ENST00000618445.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013458.hg.1 | ENST00000618445.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013458.hg.1 | ENST00000618445.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013458.hg.1 | NM_001018041.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600013458.hg.1 | NM_001018041.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600013458.hg.1 | NM_001018041.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600013458.hg.1 | NM_001018041.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600013458.hg.1 | NM_001018041.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600013458.hg.1 | NM_001018041.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600013458.hg.1 | NM_001018041.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600013458.hg.1 | NM_001018041.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600013458.hg.1 | NM_001018041.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600013458.hg.1 | NM_001018041.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600013458.hg.1 | NM_005670.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600013458.hg.1 | NM_005670.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600013458.hg.1 | NM_005670.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600013458.hg.1 | NM_005670.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600013458.hg.1 | NM_005670.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600013458.hg.1 | NM_005670.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600013458.hg.1 | NM_005670.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600013458.hg.1 | NM_005670.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600013458.hg.1 | NM_005670.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600013458.hg.1 | NM_005670.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600013458.hg.1 | NM_005670.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600013458.hg.1 | NM_005670.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0600013458.hg.1 | ENST00000367519.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013458.hg.1 | ENST00000367519.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013458.hg.1 | ENST00000367519.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013458.hg.1 | ENST00000367519.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013458.hg.1 | ENST00000367519.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013458.hg.1 | ENST00000367519.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013458.hg.1 | ENST00000367519.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013458.hg.1 | ENST00000367519.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013458.hg.1 | ENST00000367519.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013458.hg.1 | ENST00000367519.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013458.hg.1 | ENST00000367519.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013458.hg.1 | ENST00000367519.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013458.hg.1 | ENST00000367519.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013458.hg.1 | ENST00000435470.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013458.hg.1 | ENST00000435470.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013458.hg.1 | ENST00000435470.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013458.hg.1 | ENST00000435470.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013458.hg.1 | ENST00000435470.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013458.hg.1 | ENST00000435470.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013458.hg.1 | ENST00000435470.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013458.hg.1 | ENST00000450221.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013458.hg.1 | ENST00000450221.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013458.hg.1 | ENST00000450221.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013458.hg.1 | ENST00000450221.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013458.hg.1 | ENST00000450221.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013458.hg.1 | ENST00000450221.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013458.hg.1 | ENST00000461700.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013458.hg.1 | ENST00000461700.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013458.hg.1 | ENST00000461700.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013458.hg.1 | ENST00000461700.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013458.hg.1 | ENST00000461700.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013458.hg.1 | ENST00000461700.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013458.hg.1 | ENST00000461700.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013458.hg.1 | ENST00000461700.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013458.hg.1 | ENST00000489412.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013458.hg.1 | ENST00000489412.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013458.hg.1 | ENST00000489412.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013458.hg.1 | ENST00000489412.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013458.hg.1 | ENST00000496228.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013458.hg.1 | ENST00000496228.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013458.hg.1 | ENST00000496228.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013458.hg.1 | ENST00000496228.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013458.hg.1 | ENST00000496228.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013458.hg.1 | ENST00000496228.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013458.hg.1 | ENST00000496228.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013458.hg.1 | ENST00000496228.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013458.hg.1 | ENST00000496228.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013458.hg.1 | ENST00000496228.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013458.hg.1 | ENST00000611340.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013458.hg.1 | ENST00000611340.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013458.hg.1 | ENST00000611340.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013458.hg.1 | ENST00000611340.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013458.hg.1 | ENST00000611340.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013458.hg.1 | ENST00000611340.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013458.hg.1 | ENST00000611340.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013458.hg.1 | ENST00000611340.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013458.hg.1 | ENST00000611340.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013458.hg.1 | ENST00000611340.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013458.hg.1 | ENST00000618445.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013458.hg.1 | ENST00000618445.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013458.hg.1 | ENST00000618445.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013458.hg.1 | ENST00000618445.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013458.hg.1 | ENST00000618445.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013458.hg.1 | ENST00000618445.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013458.hg.1 | ENST00000618445.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013458.hg.1 | ENST00000618445.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013458.hg.1 | ENST00000618445.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013458.hg.1 | ENST00000618445.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013458.hg.1 | NM_001018041.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600013458.hg.1 | NM_001018041.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600013458.hg.1 | NM_001018041.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600013458.hg.1 | NM_001018041.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600013458.hg.1 | NM_001018041.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600013458.hg.1 | NM_001018041.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600013458.hg.1 | NM_001018041.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600013458.hg.1 | NM_001018041.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600013458.hg.1 | NM_001018041.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600013458.hg.1 | NM_001018041.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600013458.hg.1 | NM_005670.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0600013458.hg.1 | NM_005670.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0600013458.hg.1 | NM_005670.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0600013458.hg.1 | NM_005670.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0600013458.hg.1 | NM_005670.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0600013458.hg.1 | NM_005670.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0600013458.hg.1 | NM_005670.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0600013458.hg.1 | NM_005670.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0600013458.hg.1 | NM_005670.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0600013458.hg.1 | NM_005670.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0600013458.hg.1 | NM_005670.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0600013458.hg.1 | NM_005670.3 | Refseq |
