Assay Details
Gene Symbol: RAET1E | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_011535475.2 | XP_011533777.1 | 1 - 2 | 330 | 69 | |
XM_011535478.2 | XP_011533780.1 | 1 - 2 | 327 | 69 | ||
XM_011535479.2 | XP_011533781.1 | 1 - 2 | 329 | 69 | ||
XM_017010285.1 | XP_016865774.1 | 1 - 2 | 329 | 69 | ||
XM_017010286.1 | XP_016865775.1 | 1 - 2 | 329 | 69 | ||
XM_017010287.1 | XP_016865776.1 | 1 - 2 | 329 | 69 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_011535475.2, XM_011535478.2, XM_011535479.2, XM_017010285.1, XM_017010286.1, XM_017010287.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0600013536.hg.1 | ENST00000357183.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013536.hg.1 | ENST00000357183.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013536.hg.1 | ENST00000357183.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013536.hg.1 | ENST00000357183.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013536.hg.1 | ENST00000357183.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013536.hg.1 | ENST00000357183.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013536.hg.1 | ENST00000357183.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013536.hg.1 | ENST00000357183.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013536.hg.1 | ENST00000357183.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013536.hg.1 | ENST00000367363.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013536.hg.1 | ENST00000367363.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013536.hg.1 | ENST00000367363.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013536.hg.1 | ENST00000367363.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013536.hg.1 | ENST00000367363.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013536.hg.1 | ENST00000367363.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013536.hg.1 | ENST00000367363.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013536.hg.1 | ENST00000392270.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013536.hg.1 | ENST00000392270.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013536.hg.1 | ENST00000392270.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013536.hg.1 | ENST00000392270.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013536.hg.1 | ENST00000392270.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013536.hg.1 | ENST00000392270.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013536.hg.1 | ENST00000392270.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013536.hg.1 | ENST00000524905.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013536.hg.1 | ENST00000524905.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013536.hg.1 | ENST00000524905.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013536.hg.1 | ENST00000529948.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013536.hg.1 | ENST00000529948.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013536.hg.1 | ENST00000529948.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013536.hg.1 | ENST00000529948.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013536.hg.1 | ENST00000529948.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013536.hg.1 | ENST00000529948.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013536.hg.1 | ENST00000531073.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013536.hg.1 | ENST00000531073.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013536.hg.1 | ENST00000531073.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013536.hg.1 | ENST00000531073.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013536.hg.1 | ENST00000531073.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013536.hg.1 | ENST00000531073.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013536.hg.1 | ENST00000531073.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013536.hg.1 | ENST00000531073.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013536.hg.1 | ENST00000532335.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013536.hg.1 | ENST00000532335.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013536.hg.1 | ENST00000532335.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013536.hg.1 | ENST00000532335.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013536.hg.1 | ENST00000532335.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013536.hg.1 | ENST00000532335.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013536.hg.1 | ENST00000532335.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013536.hg.1 | ENST00000532335.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013536.hg.1 | ENST00000532335.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013536.hg.1 | ENST00000532335.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600013536.hg.1 | NM_001243325.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600013536.hg.1 | NM_001243325.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600013536.hg.1 | NM_001243325.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600013536.hg.1 | NM_001243325.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600013536.hg.1 | NM_001243325.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600013536.hg.1 | NM_001243325.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600013536.hg.1 | NM_001243325.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600013536.hg.1 | NM_001243325.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600013536.hg.1 | NM_001243327.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600013536.hg.1 | NM_001243327.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600013536.hg.1 | NM_001243327.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600013536.hg.1 | NM_001243327.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600013536.hg.1 | NM_001243327.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600013536.hg.1 | NM_001243327.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600013536.hg.1 | NM_001243327.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600013536.hg.1 | NM_001243327.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600013536.hg.1 | NM_001243328.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600013536.hg.1 | NM_001243328.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600013536.hg.1 | NM_001243328.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600013536.hg.1 | NM_001243328.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600013536.hg.1 | NM_001243328.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600013536.hg.1 | NM_001243328.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600013536.hg.1 | NM_001243328.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600013536.hg.1 | NM_001243328.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600013536.hg.1 | NM_001243328.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600013536.hg.1 | NM_001243328.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600013536.hg.1 | NM_139165.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600013536.hg.1 | NM_139165.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600013536.hg.1 | NM_139165.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600013536.hg.1 | NM_139165.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600013536.hg.1 | NM_139165.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600013536.hg.1 | NM_139165.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600013536.hg.1 | NM_139165.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600013536.hg.1 | NM_139165.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600013536.hg.1 | NM_139165.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600013536.hg.1 | ENST00000357183.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013536.hg.1 | ENST00000357183.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013536.hg.1 | ENST00000357183.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013536.hg.1 | ENST00000357183.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013536.hg.1 | ENST00000357183.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013536.hg.1 | ENST00000357183.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013536.hg.1 | ENST00000357183.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013536.hg.1 | ENST00000357183.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013536.hg.1 | ENST00000357183.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013536.hg.1 | ENST00000367363.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013536.hg.1 | ENST00000367363.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013536.hg.1 | ENST00000367363.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013536.hg.1 | ENST00000367363.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013536.hg.1 | ENST00000367363.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013536.hg.1 | ENST00000367363.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013536.hg.1 | ENST00000367363.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013536.hg.1 | ENST00000392270.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013536.hg.1 | ENST00000392270.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013536.hg.1 | ENST00000392270.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013536.hg.1 | ENST00000392270.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013536.hg.1 | ENST00000392270.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013536.hg.1 | ENST00000392270.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013536.hg.1 | ENST00000392270.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013536.hg.1 | ENST00000524905.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013536.hg.1 | ENST00000524905.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013536.hg.1 | ENST00000524905.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013536.hg.1 | ENST00000529948.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013536.hg.1 | ENST00000529948.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013536.hg.1 | ENST00000529948.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013536.hg.1 | ENST00000529948.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013536.hg.1 | ENST00000529948.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013536.hg.1 | ENST00000529948.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013536.hg.1 | ENST00000531073.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013536.hg.1 | ENST00000531073.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013536.hg.1 | ENST00000531073.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013536.hg.1 | ENST00000531073.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013536.hg.1 | ENST00000531073.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013536.hg.1 | ENST00000531073.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013536.hg.1 | ENST00000531073.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013536.hg.1 | ENST00000531073.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013536.hg.1 | ENST00000532335.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013536.hg.1 | ENST00000532335.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013536.hg.1 | ENST00000532335.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013536.hg.1 | ENST00000532335.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013536.hg.1 | ENST00000532335.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013536.hg.1 | ENST00000532335.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013536.hg.1 | ENST00000532335.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013536.hg.1 | ENST00000532335.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013536.hg.1 | ENST00000532335.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013536.hg.1 | ENST00000532335.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600013536.hg.1 | NM_001243325.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600013536.hg.1 | NM_001243325.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600013536.hg.1 | NM_001243325.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600013536.hg.1 | NM_001243325.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600013536.hg.1 | NM_001243325.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600013536.hg.1 | NM_001243325.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600013536.hg.1 | NM_001243325.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600013536.hg.1 | NM_001243325.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600013536.hg.1 | NM_001243327.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600013536.hg.1 | NM_001243327.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600013536.hg.1 | NM_001243327.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600013536.hg.1 | NM_001243327.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600013536.hg.1 | NM_001243327.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600013536.hg.1 | NM_001243327.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600013536.hg.1 | NM_001243327.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600013536.hg.1 | NM_001243327.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600013536.hg.1 | NM_001243328.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600013536.hg.1 | NM_001243328.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600013536.hg.1 | NM_001243328.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600013536.hg.1 | NM_001243328.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600013536.hg.1 | NM_001243328.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600013536.hg.1 | NM_001243328.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600013536.hg.1 | NM_001243328.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600013536.hg.1 | NM_001243328.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600013536.hg.1 | NM_001243328.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600013536.hg.1 | NM_001243328.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600013536.hg.1 | NM_139165.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600013536.hg.1 | NM_139165.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600013536.hg.1 | NM_139165.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600013536.hg.1 | NM_139165.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600013536.hg.1 | NM_139165.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600013536.hg.1 | NM_139165.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600013536.hg.1 | NM_139165.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600013536.hg.1 | NM_139165.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0600013536.hg.1 | NM_139165.2 | Refseq |
