Assay Details
| Gene Symbol: PREP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005267044.2 | XP_005267101.1 | 10 - 11 | 1550 | 101 | |
More Information
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0600012713.hg.1 | ENST00000369110.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012713.hg.1 | ENST00000369110.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012713.hg.1 | ENST00000369110.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012713.hg.1 | ENST00000369110.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012713.hg.1 | ENST00000369110.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012713.hg.1 | ENST00000369110.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012713.hg.1 | ENST00000369110.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012713.hg.1 | ENST00000369110.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012713.hg.1 | ENST00000369110.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012713.hg.1 | ENST00000369110.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012713.hg.1 | ENST00000369110.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012713.hg.1 | ENST00000369110.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012713.hg.1 | ENST00000369110.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012713.hg.1 | ENST00000369110.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012713.hg.1 | ENST00000369110.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012713.hg.1 | ENST00000369110.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012713.hg.1 | ENST00000369110.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012713.hg.1 | ENST00000369110.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012713.hg.1 | ENST00000448705.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012713.hg.1 | ENST00000448705.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012713.hg.1 | NM_002726.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012713.hg.1 | NM_002726.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012713.hg.1 | NM_002726.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012713.hg.1 | NM_002726.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012713.hg.1 | NM_002726.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012713.hg.1 | NM_002726.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012713.hg.1 | NM_002726.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012713.hg.1 | NM_002726.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012713.hg.1 | NM_002726.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012713.hg.1 | NM_002726.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012713.hg.1 | NM_002726.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012713.hg.1 | NM_002726.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012713.hg.1 | NM_002726.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012713.hg.1 | NM_002726.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012713.hg.1 | NM_002726.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012713.hg.1 | NM_002726.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012713.hg.1 | NM_002726.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012713.hg.1 | NM_002726.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600012713.hg.1 | NM_002726.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012713.hg.1 | ENST00000369110.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012713.hg.1 | ENST00000369110.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012713.hg.1 | ENST00000369110.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012713.hg.1 | ENST00000369110.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012713.hg.1 | ENST00000369110.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012713.hg.1 | ENST00000369110.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012713.hg.1 | ENST00000369110.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012713.hg.1 | ENST00000369110.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012713.hg.1 | ENST00000369110.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012713.hg.1 | ENST00000369110.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012713.hg.1 | ENST00000369110.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012713.hg.1 | ENST00000369110.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012713.hg.1 | ENST00000369110.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012713.hg.1 | ENST00000369110.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012713.hg.1 | ENST00000369110.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012713.hg.1 | ENST00000369110.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012713.hg.1 | ENST00000369110.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012713.hg.1 | ENST00000369110.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012713.hg.1 | ENST00000448705.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012713.hg.1 | ENST00000448705.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600012713.hg.1 | NM_002726.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012713.hg.1 | NM_002726.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012713.hg.1 | NM_002726.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012713.hg.1 | NM_002726.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012713.hg.1 | NM_002726.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012713.hg.1 | NM_002726.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012713.hg.1 | NM_002726.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012713.hg.1 | NM_002726.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012713.hg.1 | NM_002726.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012713.hg.1 | NM_002726.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012713.hg.1 | NM_002726.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012713.hg.1 | NM_002726.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012713.hg.1 | NM_002726.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012713.hg.1 | NM_002726.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012713.hg.1 | NM_002726.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012713.hg.1 | NM_002726.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012713.hg.1 | NM_002726.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012713.hg.1 | NM_002726.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600012713.hg.1 | NM_002726.4 | Refseq |

PREP
Genomic Map