Assay Details
Gene Symbol: LINC00473 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BX647106.1 | - | 1 - 2 | 518 | 104 |

More Information
Set Membership: |
![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0600014369.hg.1 | ENST00000455853.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014369.hg.1 | ENST00000455853.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014369.hg.1 | ENST00000455853.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014369.hg.1 | ENST00000455853.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014369.hg.1 | ENST00000455853.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014369.hg.1 | ENST00000581850.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014369.hg.1 | ENST00000581850.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014369.hg.1 | ENST00000581850.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014369.hg.1 | ENST00000584179.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014369.hg.1 | ENST00000584179.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014369.hg.1 | ENST00000584179.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014369.hg.1 | ENST00000584179.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014369.hg.1 | ENST00000584179.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014369.hg.1 | ENST00000584179.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014369.hg.1 | ENST00000584911.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014369.hg.1 | ENST00000584911.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014369.hg.1 | ENST00000584911.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014369.hg.1 | ENST00000584911.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014369.hg.1 | ENST00000584911.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014369.hg.1 | ENST00000584911.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014369.hg.1 | ENST00000584911.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014369.hg.1 | ENST00000584911.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014369.hg.1 | NR_026860.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600014369.hg.1 | NR_026860.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600014369.hg.1 | NR_026860.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600014369.hg.1 | NR_026860.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600014369.hg.1 | NR_026860.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600014369.hg.1 | NR_026861.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600014369.hg.1 | NR_026861.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600014369.hg.1 | NR_026861.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600014369.hg.1 | NR_026861.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600014369.hg.1 | ENST00000455853.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014369.hg.1 | ENST00000455853.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014369.hg.1 | ENST00000455853.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014369.hg.1 | ENST00000455853.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014369.hg.1 | ENST00000455853.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014369.hg.1 | ENST00000581850.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014369.hg.1 | ENST00000581850.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014369.hg.1 | ENST00000581850.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014369.hg.1 | ENST00000584179.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014369.hg.1 | ENST00000584179.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014369.hg.1 | ENST00000584179.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014369.hg.1 | ENST00000584179.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014369.hg.1 | ENST00000584179.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014369.hg.1 | ENST00000584179.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014369.hg.1 | ENST00000584911.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014369.hg.1 | ENST00000584911.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014369.hg.1 | ENST00000584911.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014369.hg.1 | ENST00000584911.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014369.hg.1 | ENST00000584911.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014369.hg.1 | ENST00000584911.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014369.hg.1 | ENST00000584911.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014369.hg.1 | ENST00000584911.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014369.hg.1 | NR_026860.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600014369.hg.1 | NR_026860.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600014369.hg.1 | NR_026860.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600014369.hg.1 | NR_026860.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600014369.hg.1 | NR_026860.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600014369.hg.1 | NR_026861.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600014369.hg.1 | NR_026861.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600014369.hg.1 | NR_026861.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600014369.hg.1 | NR_026861.1 | Refseq |
