Assay Details
| Gene Symbol: SMIM13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001135575.1 | NP_001129047.1 | 2 | 4102 | 115 | |
| GenBank mRNA | AJ420516.1 | - | 1 | 1236 | 115 | |
| AK129879.1 | - | 1 | 1055 | 115 | ||
More Information
Set Membership: |
|

SMIM13
Genomic Map