Assay Details
Gene Symbol: DHRS13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_144683.3 | NP_653284.2 | 5 | 1039 | 97 | |
GenBank mRNA | AK296017.1 | - | 3 | 690 | 97 | |
AY358566.1 | - | 5 | 1039 | 97 | ||
BC015582.2 | - | 4 | 1029 | 4637504 | 97 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1700007405.hg.1 | ENST00000579187.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007405.hg.1 | ENST00000579187.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007405.hg.1 | ENST00000579187.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007405.hg.1 | ENST00000579187.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007405.hg.1 | ENST00000580603.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007405.hg.1 | ENST00000580603.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007405.hg.1 | ENST00000580603.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007405.hg.1 | ENST00000580603.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007405.hg.1 | ENST00000580603.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007405.hg.1 | ENST00000580603.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007405.hg.1 | ENST00000580603.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007405.hg.1 | ENST00000580603.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010221.hg.1 | ENST00000378895.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010221.hg.1 | ENST00000378895.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010221.hg.1 | ENST00000378895.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010221.hg.1 | ENST00000378895.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010221.hg.1 | ENST00000378895.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010221.hg.1 | ENST00000378895.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010221.hg.1 | ENST00000378895.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010221.hg.1 | ENST00000378895.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010221.hg.1 | ENST00000378895.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010221.hg.1 | ENST00000394901.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010221.hg.1 | ENST00000394901.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010221.hg.1 | ENST00000394901.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010221.hg.1 | ENST00000394901.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010221.hg.1 | ENST00000394901.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010221.hg.1 | ENST00000394901.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010221.hg.1 | ENST00000394901.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010221.hg.1 | ENST00000394901.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010221.hg.1 | ENST00000394901.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010221.hg.1 | ENST00000394901.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010221.hg.1 | ENST00000426464.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010221.hg.1 | ENST00000426464.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010221.hg.1 | ENST00000426464.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010221.hg.1 | ENST00000426464.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010221.hg.1 | ENST00000426464.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010221.hg.1 | ENST00000426464.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010221.hg.1 | ENST00000581759.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010221.hg.1 | ENST00000581759.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010221.hg.1 | ENST00000581759.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010221.hg.1 | ENST00000581974.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010221.hg.1 | ENST00000581974.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010221.hg.1 | NM_144683.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700010221.hg.1 | NM_144683.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700010221.hg.1 | NM_144683.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700010221.hg.1 | NM_144683.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700010221.hg.1 | NM_144683.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700010221.hg.1 | NM_144683.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700010221.hg.1 | NM_144683.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700010221.hg.1 | NM_144683.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700010221.hg.1 | NM_144683.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1700007405.hg.1 | ENST00000579187.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007405.hg.1 | ENST00000579187.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007405.hg.1 | ENST00000579187.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007405.hg.1 | ENST00000579187.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007405.hg.1 | ENST00000580603.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007405.hg.1 | ENST00000580603.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007405.hg.1 | ENST00000580603.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007405.hg.1 | ENST00000580603.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007405.hg.1 | ENST00000580603.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007405.hg.1 | ENST00000580603.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007405.hg.1 | ENST00000580603.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007405.hg.1 | ENST00000580603.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010221.hg.1 | ENST00000378895.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010221.hg.1 | ENST00000378895.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010221.hg.1 | ENST00000378895.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010221.hg.1 | ENST00000378895.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010221.hg.1 | ENST00000378895.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010221.hg.1 | ENST00000378895.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010221.hg.1 | ENST00000378895.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010221.hg.1 | ENST00000378895.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010221.hg.1 | ENST00000378895.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010221.hg.1 | ENST00000394901.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010221.hg.1 | ENST00000394901.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010221.hg.1 | ENST00000394901.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010221.hg.1 | ENST00000394901.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010221.hg.1 | ENST00000394901.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010221.hg.1 | ENST00000394901.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010221.hg.1 | ENST00000394901.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010221.hg.1 | ENST00000394901.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010221.hg.1 | ENST00000394901.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010221.hg.1 | ENST00000394901.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010221.hg.1 | ENST00000426464.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010221.hg.1 | ENST00000426464.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010221.hg.1 | ENST00000426464.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010221.hg.1 | ENST00000426464.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010221.hg.1 | ENST00000426464.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010221.hg.1 | ENST00000426464.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010221.hg.1 | ENST00000581759.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010221.hg.1 | ENST00000581759.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010221.hg.1 | ENST00000581759.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010221.hg.1 | ENST00000581974.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010221.hg.1 | ENST00000581974.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010221.hg.1 | NM_144683.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1700010221.hg.1 | NM_144683.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1700010221.hg.1 | NM_144683.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1700010221.hg.1 | NM_144683.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1700010221.hg.1 | NM_144683.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1700010221.hg.1 | NM_144683.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1700010221.hg.1 | NM_144683.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1700010221.hg.1 | NM_144683.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1700010221.hg.1 | NM_144683.3 | Refseq |
