Assay Details
Gene Symbol: ERVH-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AJ431196.1 | - | 2 - 3 | 2087 | 97 | |
BC021996.1 | - | 2 - 3 | 678 | 97 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
