Assay Details
| Gene Symbol: WSB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK091514.1 | - | 1112 | 85 | ||
| Gene Symbol: INVS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC110864.1 | - | 3 | 85 | ||
More Information
Set Membership: |
|

WSB1
Genomic Map