Assay Details
Gene Symbol: WSB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK091514.1 | - | 1009 | 123 |
Gene Symbol: INVS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BC110864.1 | - | 3 | 123 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
