Assay Details
| Gene Symbol: IGFBPL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007563.2 | NP_001007564.1 | 5 | 2054 | 116 | |
| GenBank mRNA | AK055716.1 | - | 1 | 524 | 116 | |
More Information
Set Membership: |
|

IGFBPL1
Genomic Map