Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011199.hg.1 |
ENST00000302681.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011199.hg.1 |
ENST00000374428.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011199.hg.1 |
ENST00000374428.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900011199.hg.1 |
NM_205859.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900011199.hg.1 |
ENST00000302681.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011199.hg.1 |
ENST00000374428.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011199.hg.1 |
ENST00000374428.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900011199.hg.1 |
NM_205859.1 |
Refseq |