Assay Details
| Gene Symbol: HCCS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001122608.2 | NP_001116080.1 | 7 | 1863 | 105 | |
| NM_001171991.2 | NP_001165462.1 | 7 | 1826 | 105 | ||
| NM_005333.4 | NP_005324.3 | 7 | 1921 | 105 | ||
| GenBank mRNA | AI027533.1 | - | 1 | 105 | ||
| AK097815.1 | - | 7 | 1849 | 105 | ||
| BC095455.1 | - | 7 | 1864 | 30512178 | 105 | |
| CR749578.1 | - | 7 | 1878 | 105 | ||
More Information
Set Membership: |
|

HCCS
Genomic Map