Assay Details
Gene Symbol: LRCH2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001243963.1 | NP_001230892.1 | 20 | 3661 | 111 | |
NM_020871.3 | NP_065922.3 | 21 | 3712 | 111 | ||
XM_006724724.3 | XP_006724787.2 | 21 | 3694 | 111 | ||
GenBank mRNA | AB040928.1 | - | 17 | 2893 | 111 | |
AK022128.1 | - | 13 | 2468 | 111 | ||
BC042004.1 | - | 1 | 1449 | 110 | ||
BC125223.1 | - | 9 | 2119 | 111 | ||
BC125224.1 | - | 20 | 3654 | 40124466 | 111 | |
BX647193.1 | - | 21 | 3712 | 110 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | ENST00000317135.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | ENST00000317135.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | ENST00000317135.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | ENST00000317135.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | ENST00000317135.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | ENST00000317135.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | ENST00000317135.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | ENST00000317135.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | ENST00000317135.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | ENST00000317135.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | ENST00000317135.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | ENST00000317135.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | ENST00000317135.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | ENST00000317135.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | ENST00000317135.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | ENST00000317135.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | ENST00000317135.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | ENST00000317135.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | ENST00000317135.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | ENST00000317135.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | ENST00000317135.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | ENST00000317135.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | ENST00000317135.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | ENST00000317135.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | ENST00000317135.12 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | ENST00000538422.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | ENST00000538422.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | ENST00000538422.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | ENST00000538422.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | ENST00000538422.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | ENST00000538422.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | ENST00000538422.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | ENST00000538422.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | ENST00000538422.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | ENST00000538422.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | ENST00000538422.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | ENST00000538422.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | ENST00000538422.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | ENST00000538422.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | ENST00000538422.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | ENST00000538422.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | ENST00000538422.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | ENST00000538422.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | ENST00000538422.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | ENST00000538422.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | ENST00000538422.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | ENST00000538422.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | ENST00000538422.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | ENST00000538422.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | ENST00000607417.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | NM_001243963.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | NM_001243963.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | NM_001243963.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | NM_001243963.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | NM_001243963.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | NM_001243963.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | NM_001243963.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | NM_001243963.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | NM_001243963.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | NM_001243963.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | NM_001243963.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | NM_001243963.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | NM_001243963.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | NM_001243963.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | NM_001243963.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | NM_001243963.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | NM_001243963.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | NM_001243963.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | NM_001243963.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | NM_001243963.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | NM_001243963.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | NM_001243963.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | NM_001243963.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | NM_001243963.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | NM_020871.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | NM_020871.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | NM_020871.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | NM_020871.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | NM_020871.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | NM_020871.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | NM_020871.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | NM_020871.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | NM_020871.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | NM_020871.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | NM_020871.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | NM_020871.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | NM_020871.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | NM_020871.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | NM_020871.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | NM_020871.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | NM_020871.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | NM_020871.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | NM_020871.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | NM_020871.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | NM_020871.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | NM_020871.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | NM_020871.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | NM_020871.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00010559.hg.1 | NM_020871.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | ENST00000317135.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | ENST00000317135.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | ENST00000317135.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | ENST00000317135.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | ENST00000317135.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | ENST00000317135.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | ENST00000317135.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | ENST00000317135.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | ENST00000317135.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | ENST00000317135.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | ENST00000317135.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | ENST00000317135.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | ENST00000317135.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | ENST00000317135.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | ENST00000317135.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | ENST00000317135.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | ENST00000317135.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | ENST00000317135.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | ENST00000317135.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | ENST00000317135.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | ENST00000317135.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | ENST00000317135.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | ENST00000317135.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | ENST00000317135.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | ENST00000317135.12 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | ENST00000538422.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | ENST00000538422.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | ENST00000538422.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | ENST00000538422.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | ENST00000538422.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | ENST00000538422.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | ENST00000538422.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | ENST00000538422.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | ENST00000538422.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | ENST00000538422.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | ENST00000538422.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | ENST00000538422.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | ENST00000538422.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | ENST00000538422.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | ENST00000538422.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | ENST00000538422.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | ENST00000538422.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | ENST00000538422.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | ENST00000538422.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | ENST00000538422.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | ENST00000538422.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | ENST00000538422.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | ENST00000538422.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | ENST00000538422.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | ENST00000607417.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | NM_001243963.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | NM_001243963.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | NM_001243963.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | NM_001243963.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | NM_001243963.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | NM_001243963.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | NM_001243963.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | NM_001243963.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | NM_001243963.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | NM_001243963.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | NM_001243963.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | NM_001243963.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | NM_001243963.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | NM_001243963.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | NM_001243963.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | NM_001243963.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | NM_001243963.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | NM_001243963.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | NM_001243963.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | NM_001243963.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | NM_001243963.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | NM_001243963.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | NM_001243963.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | NM_001243963.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | NM_020871.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | NM_020871.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | NM_020871.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | NM_020871.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | NM_020871.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | NM_020871.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | NM_020871.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | NM_020871.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | NM_020871.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | NM_020871.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | NM_020871.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | NM_020871.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | NM_020871.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | NM_020871.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | NM_020871.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | NM_020871.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | NM_020871.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | NM_020871.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | NM_020871.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | NM_020871.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | NM_020871.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | NM_020871.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | NM_020871.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | NM_020871.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00010559.hg.1 | NM_020871.3 | Refseq |
