Assay Details
| Gene Symbol: FLJ40194 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK097513.1 | - | 2 - 3 | 493 | 68 | |
More Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1700008196.hg.1 | ENST00000322227.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008196.hg.1 | ENST00000322227.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008196.hg.1 | ENST00000322227.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008196.hg.1 | ENST00000322227.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008196.hg.1 | ENST00000322227.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008196.hg.1 | ENST00000322227.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008196.hg.1 | ENST00000322227.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008196.hg.1 | ENST00000322227.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008196.hg.1 | ENST00000322227.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008196.hg.1 | ENST00000322227.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008196.hg.1 | ENST00000322227.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008196.hg.1 | ENST00000322227.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008196.hg.1 | ENST00000322227.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008196.hg.1 | ENST00000322227.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008196.hg.1 | ENST00000322227.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008196.hg.1 | ENST00000322227.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008196.hg.1 | ENST00000322227.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008196.hg.1 | ENST00000322227.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008196.hg.1 | ENST00000322227.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008196.hg.1 | ENST00000322227.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008196.hg.1 | ENST00000322227.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008196.hg.1 | ENST00000322227.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008196.hg.1 | ENST00000322227.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008196.hg.1 | ENST00000322227.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008196.hg.1 | ENST00000502951.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008196.hg.1 | ENST00000502951.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008196.hg.1 | ENST00000502951.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008196.hg.1 | ENST00000511008.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008196.hg.1 | ENST00000511008.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008196.hg.1 | ENST00000511008.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008196.hg.1 | ENST00000511008.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008196.hg.1 | ENST00000511008.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008196.hg.1 | NR_034161.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008196.hg.1 | NR_034161.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008196.hg.1 | NR_034161.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008196.hg.1 | NR_034161.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008196.hg.1 | NR_034161.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008196.hg.1 | ENST00000322227.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008196.hg.1 | ENST00000322227.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008196.hg.1 | ENST00000322227.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008196.hg.1 | ENST00000322227.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008196.hg.1 | ENST00000322227.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008196.hg.1 | ENST00000322227.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008196.hg.1 | ENST00000322227.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008196.hg.1 | ENST00000322227.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008196.hg.1 | ENST00000322227.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008196.hg.1 | ENST00000322227.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008196.hg.1 | ENST00000322227.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008196.hg.1 | ENST00000322227.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008196.hg.1 | ENST00000322227.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008196.hg.1 | ENST00000322227.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008196.hg.1 | ENST00000322227.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008196.hg.1 | ENST00000322227.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008196.hg.1 | ENST00000322227.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008196.hg.1 | ENST00000322227.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008196.hg.1 | ENST00000322227.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008196.hg.1 | ENST00000322227.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008196.hg.1 | ENST00000322227.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008196.hg.1 | ENST00000322227.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008196.hg.1 | ENST00000322227.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008196.hg.1 | ENST00000322227.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008196.hg.1 | ENST00000502951.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008196.hg.1 | ENST00000502951.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008196.hg.1 | ENST00000502951.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008196.hg.1 | ENST00000511008.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008196.hg.1 | ENST00000511008.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008196.hg.1 | ENST00000511008.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008196.hg.1 | ENST00000511008.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008196.hg.1 | ENST00000511008.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008196.hg.1 | NR_034161.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008196.hg.1 | NR_034161.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008196.hg.1 | NR_034161.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008196.hg.1 | NR_034161.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008196.hg.1 | NR_034161.1 | Refseq |

FLJ40194
Genomic Map