Assay Details
Gene Symbol: NACC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005259721.3 | XP_005259778.1 | 1 - 2 | 87 | 71 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005259721.3 transcripts.
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1900007125.hg.1 | ENST00000292431.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007125.hg.1 | ENST00000292431.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007125.hg.1 | ENST00000292431.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007125.hg.1 | ENST00000292431.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007125.hg.1 | ENST00000292431.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007125.hg.1 | ENST00000292431.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007125.hg.1 | ENST00000292431.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007125.hg.1 | ENST00000292431.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007125.hg.1 | ENST00000292431.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007125.hg.1 | ENST00000292431.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007125.hg.1 | ENST00000292431.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007125.hg.1 | ENST00000292431.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007125.hg.1 | ENST00000585663.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007125.hg.1 | ENST00000585663.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007125.hg.1 | ENST00000585663.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007125.hg.1 | ENST00000586171.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007125.hg.1 | ENST00000586171.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900007125.hg.1 | NM_052876.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900007125.hg.1 | NM_052876.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900007125.hg.1 | NM_052876.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900007125.hg.1 | NM_052876.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900007125.hg.1 | NM_052876.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900007125.hg.1 | NM_052876.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900007125.hg.1 | NM_052876.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900007125.hg.1 | NM_052876.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900007125.hg.1 | NM_052876.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900007125.hg.1 | NM_052876.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900007125.hg.1 | NM_052876.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900007125.hg.1 | NM_052876.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900007125.hg.1 | NM_052876.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1900007125.hg.1 | ENST00000292431.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007125.hg.1 | ENST00000292431.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007125.hg.1 | ENST00000292431.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007125.hg.1 | ENST00000292431.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007125.hg.1 | ENST00000292431.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007125.hg.1 | ENST00000292431.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007125.hg.1 | ENST00000292431.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007125.hg.1 | ENST00000292431.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007125.hg.1 | ENST00000292431.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007125.hg.1 | ENST00000292431.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007125.hg.1 | ENST00000292431.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007125.hg.1 | ENST00000292431.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007125.hg.1 | ENST00000585663.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007125.hg.1 | ENST00000585663.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007125.hg.1 | ENST00000585663.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007125.hg.1 | ENST00000586171.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007125.hg.1 | ENST00000586171.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900007125.hg.1 | NM_052876.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1900007125.hg.1 | NM_052876.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1900007125.hg.1 | NM_052876.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1900007125.hg.1 | NM_052876.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1900007125.hg.1 | NM_052876.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1900007125.hg.1 | NM_052876.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1900007125.hg.1 | NM_052876.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1900007125.hg.1 | NM_052876.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1900007125.hg.1 | NM_052876.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1900007125.hg.1 | NM_052876.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1900007125.hg.1 | NM_052876.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1900007125.hg.1 | NM_052876.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1900007125.hg.1 | NM_052876.3 | Refseq |
