Assay Details
| Gene Symbol: LOC102724958 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_110760.1 | 3 - 4 | 812 | 84 | ||
| GenBank mRNA | BX092781.1 | - | 3 - 4 | 383 | 84 | |
More Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1900007659.hg.1 | ENST00000585813.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007659.hg.1 | ENST00000585813.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007659.hg.1 | ENST00000585813.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007659.hg.1 | ENST00000585813.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007659.hg.1 | ENST00000585813.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007659.hg.1 | ENST00000585813.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007659.hg.1 | ENST00000585813.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007659.hg.1 | ENST00000585813.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007659.hg.1 | ENST00000585813.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007659.hg.1 | ENST00000585813.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007659.hg.1 | ENST00000585813.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007659.hg.1 | ENST00000585813.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007659.hg.1 | ENST00000585813.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007659.hg.1 | ENST00000585813.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007659.hg.1 | ENST00000587998.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007659.hg.1 | ENST00000587998.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007659.hg.1 | ENST00000587998.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007659.hg.1 | ENST00000587998.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007659.hg.1 | ENST00000587998.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007659.hg.1 | ENST00000587998.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007659.hg.1 | ENST00000587998.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007659.hg.1 | ENST00000587998.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007659.hg.1 | ENST00000587998.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007659.hg.1 | ENST00000587998.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007659.hg.1 | ENST00000587998.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007659.hg.1 | ENST00000587998.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007659.hg.1 | ENST00000587998.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007659.hg.1 | ENST00000587998.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007659.hg.1 | ENST00000587998.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007659.hg.1 | ENST00000587998.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007659.hg.1 | ENST00000587998.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007659.hg.1 | ENST00000587998.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007659.hg.1 | ENST00000588609.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007659.hg.1 | ENST00000588609.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007659.hg.1 | ENST00000588609.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007659.hg.1 | ENST00000588609.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007659.hg.1 | ENST00000588609.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007659.hg.1 | ENST00000589466.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007659.hg.1 | ENST00000589466.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007659.hg.1 | ENST00000589466.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007659.hg.1 | ENST00000589466.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007659.hg.1 | ENST00000589466.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007659.hg.1 | ENST00000589466.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007659.hg.1 | ENST00000589623.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007659.hg.1 | ENST00000589623.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007659.hg.1 | ENST00000589623.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007659.hg.1 | ENST00000589623.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007659.hg.1 | ENST00000589921.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007659.hg.1 | ENST00000589921.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007659.hg.1 | ENST00000589921.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007659.hg.1 | ENST00000589921.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007659.hg.1 | ENST00000589921.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007659.hg.1 | ENST00000589921.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007659.hg.1 | ENST00000589921.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007659.hg.1 | ENST00000589921.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007659.hg.1 | ENST00000589921.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007659.hg.1 | ENST00000589921.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007659.hg.1 | ENST00000589921.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007659.hg.1 | NR_027318.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007659.hg.1 | NR_027318.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007659.hg.1 | NR_027318.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007659.hg.1 | NR_027318.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007659.hg.1 | NR_027318.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007659.hg.1 | NR_027318.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007659.hg.1 | NR_027318.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007659.hg.1 | NR_027318.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007659.hg.1 | NR_027318.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007659.hg.1 | NR_027318.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007659.hg.1 | NR_027318.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007659.hg.1 | NR_110760.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007659.hg.1 | NR_110760.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007659.hg.1 | NR_110760.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007659.hg.1 | NR_110760.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007659.hg.1 | NR_110760.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007659.hg.1 | NR_110760.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007659.hg.1 | NR_110760.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007659.hg.1 | NR_110760.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007659.hg.1 | NR_110760.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900007659.hg.1 | NR_110760.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007659.hg.1 | ENST00000585813.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007659.hg.1 | ENST00000585813.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007659.hg.1 | ENST00000585813.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007659.hg.1 | ENST00000585813.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007659.hg.1 | ENST00000585813.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007659.hg.1 | ENST00000585813.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007659.hg.1 | ENST00000585813.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007659.hg.1 | ENST00000585813.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007659.hg.1 | ENST00000585813.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007659.hg.1 | ENST00000585813.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007659.hg.1 | ENST00000585813.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007659.hg.1 | ENST00000585813.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007659.hg.1 | ENST00000585813.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007659.hg.1 | ENST00000585813.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007659.hg.1 | ENST00000587998.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007659.hg.1 | ENST00000587998.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007659.hg.1 | ENST00000587998.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007659.hg.1 | ENST00000587998.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007659.hg.1 | ENST00000587998.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007659.hg.1 | ENST00000587998.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007659.hg.1 | ENST00000587998.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007659.hg.1 | ENST00000587998.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007659.hg.1 | ENST00000587998.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007659.hg.1 | ENST00000587998.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007659.hg.1 | ENST00000587998.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007659.hg.1 | ENST00000587998.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007659.hg.1 | ENST00000587998.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007659.hg.1 | ENST00000587998.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007659.hg.1 | ENST00000587998.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007659.hg.1 | ENST00000587998.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007659.hg.1 | ENST00000587998.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007659.hg.1 | ENST00000587998.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007659.hg.1 | ENST00000588609.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007659.hg.1 | ENST00000588609.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007659.hg.1 | ENST00000588609.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007659.hg.1 | ENST00000588609.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007659.hg.1 | ENST00000588609.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007659.hg.1 | ENST00000589466.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007659.hg.1 | ENST00000589466.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007659.hg.1 | ENST00000589466.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007659.hg.1 | ENST00000589466.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007659.hg.1 | ENST00000589466.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007659.hg.1 | ENST00000589466.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007659.hg.1 | ENST00000589623.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007659.hg.1 | ENST00000589623.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007659.hg.1 | ENST00000589623.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007659.hg.1 | ENST00000589623.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007659.hg.1 | ENST00000589921.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007659.hg.1 | ENST00000589921.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007659.hg.1 | ENST00000589921.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007659.hg.1 | ENST00000589921.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007659.hg.1 | ENST00000589921.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007659.hg.1 | ENST00000589921.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007659.hg.1 | ENST00000589921.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007659.hg.1 | ENST00000589921.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007659.hg.1 | ENST00000589921.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007659.hg.1 | ENST00000589921.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007659.hg.1 | ENST00000589921.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900007659.hg.1 | NR_027318.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007659.hg.1 | NR_027318.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007659.hg.1 | NR_027318.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007659.hg.1 | NR_027318.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007659.hg.1 | NR_027318.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007659.hg.1 | NR_027318.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007659.hg.1 | NR_027318.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007659.hg.1 | NR_027318.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007659.hg.1 | NR_027318.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007659.hg.1 | NR_027318.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007659.hg.1 | NR_027318.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007659.hg.1 | NR_110760.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007659.hg.1 | NR_110760.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007659.hg.1 | NR_110760.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007659.hg.1 | NR_110760.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007659.hg.1 | NR_110760.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007659.hg.1 | NR_110760.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007659.hg.1 | NR_110760.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007659.hg.1 | NR_110760.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007659.hg.1 | NR_110760.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900007659.hg.1 | NR_110760.1 | Refseq |

LOC102724958
Genomic Map