Assay Details
Gene Symbol: CENPBD1P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BC065480.1 | - | 1 - 2 | 149 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1900011832.hg.1 | ENST00000464061.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011832.hg.1 | ENST00000473164.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011832.hg.1 | ENST00000473164.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011832.hg.1 | ENST00000473164.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011832.hg.1 | ENST00000473164.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011832.hg.1 | ENST00000473164.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011832.hg.1 | ENST00000473164.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011832.hg.1 | ENST00000473164.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011832.hg.1 | ENST00000487264.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011832.hg.1 | ENST00000487264.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011832.hg.1 | ENST00000487264.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011832.hg.1 | ENST00000487264.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011832.hg.1 | ENST00000487264.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011832.hg.1 | ENST00000487264.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011832.hg.1 | ENST00000487264.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011832.hg.1 | ENST00000487264.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011832.hg.1 | ENST00000487264.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011832.hg.1 | ENST00000487264.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011832.hg.1 | ENST00000492393.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011832.hg.1 | ENST00000492393.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011832.hg.1 | ENST00000492393.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011832.hg.1 | ENST00000492393.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011832.hg.1 | ENST00000493504.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011832.hg.1 | ENST00000493504.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011832.hg.1 | ENST00000493504.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011832.hg.1 | ENST00000493504.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011832.hg.1 | ENST00000493504.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011832.hg.1 | ENST00000493504.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011832.hg.1 | ENST00000493504.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011832.hg.1 | ENST00000595155.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011832.hg.1 | ENST00000595155.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011832.hg.1 | ENST00000595155.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011832.hg.1 | ENST00000595439.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011832.hg.1 | ENST00000595439.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011832.hg.1 | ENST00000595439.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011832.hg.1 | ENST00000595439.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011832.hg.1 | ENST00000596579.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011832.hg.1 | ENST00000596579.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011832.hg.1 | ENST00000597341.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011832.hg.1 | ENST00000597341.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900011832.hg.1 | NR_026052.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900011832.hg.1 | NR_026052.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900011832.hg.1 | NR_026052.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900011832.hg.1 | NR_026052.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900011832.hg.1 | NR_026052.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900011832.hg.1 | NR_026052.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900011832.hg.1 | NR_026052.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1900011832.hg.1 | NR_026052.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900009330.hg.1 | ENST00000442569.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009330.hg.1 | ENST00000442569.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009330.hg.1 | ENST00000442569.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009330.hg.1 | ENST00000442569.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009330.hg.1 | ENST00000442569.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009330.hg.1 | ENST00000442569.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009330.hg.1 | ENST00000442569.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900009330.hg.1 | ENST00000442569.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011832.hg.1 | ENST00000464061.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011832.hg.1 | ENST00000473164.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011832.hg.1 | ENST00000473164.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011832.hg.1 | ENST00000473164.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011832.hg.1 | ENST00000473164.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011832.hg.1 | ENST00000473164.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011832.hg.1 | ENST00000473164.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011832.hg.1 | ENST00000473164.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011832.hg.1 | ENST00000487264.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011832.hg.1 | ENST00000487264.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011832.hg.1 | ENST00000487264.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011832.hg.1 | ENST00000487264.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011832.hg.1 | ENST00000487264.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011832.hg.1 | ENST00000487264.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011832.hg.1 | ENST00000487264.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011832.hg.1 | ENST00000487264.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011832.hg.1 | ENST00000487264.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011832.hg.1 | ENST00000487264.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011832.hg.1 | ENST00000492393.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011832.hg.1 | ENST00000492393.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011832.hg.1 | ENST00000492393.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011832.hg.1 | ENST00000492393.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011832.hg.1 | ENST00000493504.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011832.hg.1 | ENST00000493504.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011832.hg.1 | ENST00000493504.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011832.hg.1 | ENST00000493504.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011832.hg.1 | ENST00000493504.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011832.hg.1 | ENST00000493504.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011832.hg.1 | ENST00000493504.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011832.hg.1 | ENST00000595155.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011832.hg.1 | ENST00000595155.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011832.hg.1 | ENST00000595155.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011832.hg.1 | ENST00000595439.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011832.hg.1 | ENST00000595439.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011832.hg.1 | ENST00000595439.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011832.hg.1 | ENST00000595439.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011832.hg.1 | ENST00000596579.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011832.hg.1 | ENST00000596579.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011832.hg.1 | ENST00000597341.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011832.hg.1 | ENST00000597341.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900011832.hg.1 | NR_026052.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900011832.hg.1 | NR_026052.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900011832.hg.1 | NR_026052.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900011832.hg.1 | NR_026052.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900011832.hg.1 | NR_026052.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900011832.hg.1 | NR_026052.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900011832.hg.1 | NR_026052.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1900011832.hg.1 | NR_026052.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900009330.hg.1 | ENST00000442569.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009330.hg.1 | ENST00000442569.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009330.hg.1 | ENST00000442569.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009330.hg.1 | ENST00000442569.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009330.hg.1 | ENST00000442569.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009330.hg.1 | ENST00000442569.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009330.hg.1 | ENST00000442569.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900009330.hg.1 | ENST00000442569.1 | Ensembl |
