Assay Details
Gene Symbol: KCND3-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BX098424.1 | - | 1 - 2 | 196 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0100009431.hg.1 | ENST00000419258.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009431.hg.1 | ENST00000419258.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009431.hg.1 | ENST00000419258.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009431.hg.1 | ENST00000419258.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009431.hg.1 | ENST00000419258.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009431.hg.1 | ENST00000419258.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009431.hg.1 | ENST00000427471.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009431.hg.1 | ENST00000427471.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009431.hg.1 | ENST00000427471.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009431.hg.1 | ENST00000427471.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009431.hg.1 | ENST00000427471.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009431.hg.1 | ENST00000427471.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009431.hg.1 | ENST00000427471.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009431.hg.1 | ENST00000427471.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009431.hg.1 | ENST00000427471.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009431.hg.1 | ENST00000441125.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009431.hg.1 | ENST00000441125.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009431.hg.1 | ENST00000441125.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009431.hg.1 | ENST00000441125.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009431.hg.1 | ENST00000441125.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009431.hg.1 | ENST00000441125.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009431.hg.1 | NR_046619.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009431.hg.1 | NR_046619.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009431.hg.1 | NR_046619.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009431.hg.1 | NR_046619.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009431.hg.1 | NR_046619.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009431.hg.1 | NR_046619.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009431.hg.1 | NR_046619.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009431.hg.1 | NR_046619.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009431.hg.1 | ENST00000419258.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009431.hg.1 | ENST00000419258.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009431.hg.1 | ENST00000419258.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009431.hg.1 | ENST00000419258.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009431.hg.1 | ENST00000419258.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009431.hg.1 | ENST00000419258.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009431.hg.1 | ENST00000427471.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009431.hg.1 | ENST00000427471.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009431.hg.1 | ENST00000427471.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009431.hg.1 | ENST00000427471.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009431.hg.1 | ENST00000427471.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009431.hg.1 | ENST00000427471.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009431.hg.1 | ENST00000427471.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009431.hg.1 | ENST00000427471.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009431.hg.1 | ENST00000427471.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009431.hg.1 | ENST00000441125.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009431.hg.1 | ENST00000441125.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009431.hg.1 | ENST00000441125.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009431.hg.1 | ENST00000441125.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009431.hg.1 | ENST00000441125.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009431.hg.1 | ENST00000441125.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009431.hg.1 | NR_046619.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009431.hg.1 | NR_046619.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009431.hg.1 | NR_046619.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009431.hg.1 | NR_046619.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009431.hg.1 | NR_046619.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009431.hg.1 | NR_046619.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009431.hg.1 | NR_046619.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009431.hg.1 | NR_046619.1 | Refseq |
