Assay Details
Gene Symbol: C1orf105 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_011510152.2 | XP_011508454.1 | 1 - 2 | 2349 | 120 | |
XM_011510155.2 | XP_011508457.1 | 1 - 2 | 2349 | 120 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_011510152.2, XM_011510155.2 transcripts.
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0100010621.hg.1 | ENST00000367725.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010621.hg.1 | ENST00000367725.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010621.hg.1 | ENST00000367725.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010621.hg.1 | ENST00000367725.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010621.hg.1 | ENST00000367725.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010621.hg.1 | ENST00000367725.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010621.hg.1 | ENST00000367725.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010621.hg.1 | ENST00000367726.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010621.hg.1 | ENST00000367726.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010621.hg.1 | ENST00000367726.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010621.hg.1 | ENST00000367726.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010621.hg.1 | ENST00000367726.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010621.hg.1 | ENST00000367727.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010621.hg.1 | ENST00000367727.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010621.hg.1 | ENST00000367727.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010621.hg.1 | ENST00000367727.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010621.hg.1 | ENST00000367727.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010621.hg.1 | ENST00000367727.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010621.hg.1 | ENST00000367727.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010621.hg.1 | ENST00000367727.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010621.hg.1 | ENST00000367727.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010621.hg.1 | ENST00000460397.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010621.hg.1 | ENST00000460397.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010621.hg.1 | ENST00000460397.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010621.hg.1 | ENST00000460397.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010621.hg.1 | ENST00000488100.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010621.hg.1 | ENST00000488100.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010621.hg.1 | ENST00000488100.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010621.hg.1 | ENST00000488100.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010621.hg.1 | ENST00000488100.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010621.hg.1 | NM_001300760.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100010621.hg.1 | NM_001300760.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100010621.hg.1 | NM_001300760.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100010621.hg.1 | NM_001300760.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100010621.hg.1 | NM_001300760.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100010621.hg.1 | NM_001300760.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100010621.hg.1 | NM_001300760.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100010621.hg.1 | NM_139240.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100010621.hg.1 | NM_139240.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100010621.hg.1 | NM_139240.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100010621.hg.1 | NM_139240.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100010621.hg.1 | NM_139240.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100010621.hg.1 | NM_139240.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100010621.hg.1 | NM_139240.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100010621.hg.1 | NM_139240.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100010621.hg.1 | NM_139240.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100010621.hg.1 | ENST00000367725.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010621.hg.1 | ENST00000367725.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010621.hg.1 | ENST00000367725.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010621.hg.1 | ENST00000367725.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010621.hg.1 | ENST00000367725.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010621.hg.1 | ENST00000367725.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010621.hg.1 | ENST00000367725.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010621.hg.1 | ENST00000367726.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010621.hg.1 | ENST00000367726.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010621.hg.1 | ENST00000367726.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010621.hg.1 | ENST00000367726.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010621.hg.1 | ENST00000367726.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010621.hg.1 | ENST00000367727.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010621.hg.1 | ENST00000367727.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010621.hg.1 | ENST00000367727.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010621.hg.1 | ENST00000367727.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010621.hg.1 | ENST00000367727.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010621.hg.1 | ENST00000367727.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010621.hg.1 | ENST00000367727.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010621.hg.1 | ENST00000367727.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010621.hg.1 | ENST00000367727.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010621.hg.1 | ENST00000460397.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010621.hg.1 | ENST00000460397.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010621.hg.1 | ENST00000460397.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010621.hg.1 | ENST00000460397.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010621.hg.1 | ENST00000488100.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010621.hg.1 | ENST00000488100.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010621.hg.1 | ENST00000488100.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010621.hg.1 | ENST00000488100.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010621.hg.1 | ENST00000488100.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010621.hg.1 | NM_001300760.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100010621.hg.1 | NM_001300760.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100010621.hg.1 | NM_001300760.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100010621.hg.1 | NM_001300760.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100010621.hg.1 | NM_001300760.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100010621.hg.1 | NM_001300760.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100010621.hg.1 | NM_001300760.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100010621.hg.1 | NM_139240.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100010621.hg.1 | NM_139240.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100010621.hg.1 | NM_139240.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100010621.hg.1 | NM_139240.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100010621.hg.1 | NM_139240.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100010621.hg.1 | NM_139240.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100010621.hg.1 | NM_139240.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100010621.hg.1 | NM_139240.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100010621.hg.1 | NM_139240.3 | Refseq |
