Assay Details
| Gene Symbol: HOOK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_015888.4 | NP_056972.1 | 21 - 22 | 2276 | 115 | |
| XM_011541562.2 | XP_011539864.1 | 21 - 22 | 2246 | 115 | ||
| GenBank mRNA | AF044923.1 | - | 21 - 22 | 2218 | 115 | |
| AK097735.1 | - | 19 - 20 | 2019 | 115 | ||
| AK098561.1 | - | 15 - 16 | 1557 | 115 | ||
| AK292314.1 | - | 22 - 23 | 2121 | 115 | ||
| AK301768.1 | - | 21 - 22 | 2044 | 115 | ||
| BC011621.2 | - | 21 - 22 | 2276 | 3959931 | 115 | |
More Information
Set Membership: |
|

HOOK1
Genomic Map