Assay Details
Gene Symbol: SARS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_034072.1 | 1 - 2 | 243 | 92 | ||
GenBank mRNA | DA792457.1 | - | 1 - 2 | 232 | 92 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0100009344.hg.1 | ENST00000234677.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009344.hg.1 | ENST00000234677.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009344.hg.1 | ENST00000234677.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009344.hg.1 | ENST00000234677.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009344.hg.1 | ENST00000234677.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009344.hg.1 | ENST00000234677.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009344.hg.1 | ENST00000234677.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009344.hg.1 | ENST00000234677.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009344.hg.1 | ENST00000234677.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009344.hg.1 | ENST00000234677.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009344.hg.1 | ENST00000234677.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009344.hg.1 | ENST00000234677.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009344.hg.1 | ENST00000369923.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009344.hg.1 | ENST00000369923.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009344.hg.1 | ENST00000369923.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009344.hg.1 | ENST00000369923.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009344.hg.1 | ENST00000369923.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009344.hg.1 | ENST00000369923.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009344.hg.1 | ENST00000369923.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009344.hg.1 | ENST00000369923.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009344.hg.1 | ENST00000369923.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009344.hg.1 | ENST00000369923.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009344.hg.1 | ENST00000369923.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009344.hg.1 | ENST00000369923.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009344.hg.1 | ENST00000369923.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009344.hg.1 | ENST00000369923.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009344.hg.1 | ENST00000468588.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009344.hg.1 | ENST00000468588.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009344.hg.1 | ENST00000468588.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009344.hg.1 | ENST00000468588.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009344.hg.1 | ENST00000468588.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009344.hg.1 | ENST00000468588.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009344.hg.1 | ENST00000468588.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009344.hg.1 | ENST00000468588.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009344.hg.1 | ENST00000468588.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009344.hg.1 | ENST00000471705.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009344.hg.1 | ENST00000471705.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009344.hg.1 | ENST00000471705.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009344.hg.1 | ENST00000477544.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009344.hg.1 | ENST00000477544.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009344.hg.1 | ENST00000477544.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009344.hg.1 | ENST00000477544.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009344.hg.1 | ENST00000477544.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009344.hg.1 | ENST00000482384.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009344.hg.1 | ENST00000482384.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009344.hg.1 | NM_006513.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009344.hg.1 | NM_006513.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009344.hg.1 | NM_006513.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009344.hg.1 | NM_006513.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009344.hg.1 | NM_006513.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009344.hg.1 | NM_006513.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009344.hg.1 | NM_006513.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009344.hg.1 | NM_006513.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009344.hg.1 | NM_006513.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009344.hg.1 | NM_006513.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009344.hg.1 | NM_006513.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009344.hg.1 | NM_006513.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009344.hg.1 | NM_006513.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009344.hg.1 | NM_006513.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009344.hg.1 | NR_034072.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009344.hg.1 | NR_034072.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009344.hg.1 | NR_034072.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009344.hg.1 | NR_034072.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100010826.hg.1 | ENST00000621054.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010826.hg.1 | ENST00000621054.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009344.hg.1 | ENST00000234677.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009344.hg.1 | ENST00000234677.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009344.hg.1 | ENST00000234677.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009344.hg.1 | ENST00000234677.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009344.hg.1 | ENST00000234677.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009344.hg.1 | ENST00000234677.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009344.hg.1 | ENST00000234677.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009344.hg.1 | ENST00000234677.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009344.hg.1 | ENST00000234677.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009344.hg.1 | ENST00000234677.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009344.hg.1 | ENST00000234677.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009344.hg.1 | ENST00000234677.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009344.hg.1 | ENST00000369923.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009344.hg.1 | ENST00000369923.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009344.hg.1 | ENST00000369923.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009344.hg.1 | ENST00000369923.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009344.hg.1 | ENST00000369923.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009344.hg.1 | ENST00000369923.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009344.hg.1 | ENST00000369923.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009344.hg.1 | ENST00000369923.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009344.hg.1 | ENST00000369923.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009344.hg.1 | ENST00000369923.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009344.hg.1 | ENST00000369923.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009344.hg.1 | ENST00000369923.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009344.hg.1 | ENST00000369923.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009344.hg.1 | ENST00000369923.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009344.hg.1 | ENST00000468588.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009344.hg.1 | ENST00000468588.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009344.hg.1 | ENST00000468588.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009344.hg.1 | ENST00000468588.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009344.hg.1 | ENST00000468588.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009344.hg.1 | ENST00000468588.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009344.hg.1 | ENST00000468588.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009344.hg.1 | ENST00000468588.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009344.hg.1 | ENST00000468588.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009344.hg.1 | ENST00000471705.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009344.hg.1 | ENST00000471705.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009344.hg.1 | ENST00000471705.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009344.hg.1 | ENST00000477544.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009344.hg.1 | ENST00000477544.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009344.hg.1 | ENST00000477544.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009344.hg.1 | ENST00000477544.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009344.hg.1 | ENST00000477544.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009344.hg.1 | ENST00000482384.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009344.hg.1 | ENST00000482384.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009344.hg.1 | NM_006513.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009344.hg.1 | NM_006513.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009344.hg.1 | NM_006513.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009344.hg.1 | NM_006513.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009344.hg.1 | NM_006513.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009344.hg.1 | NM_006513.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009344.hg.1 | NM_006513.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009344.hg.1 | NM_006513.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009344.hg.1 | NM_006513.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009344.hg.1 | NM_006513.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009344.hg.1 | NM_006513.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009344.hg.1 | NM_006513.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009344.hg.1 | NM_006513.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009344.hg.1 | NM_006513.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009344.hg.1 | NR_034072.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009344.hg.1 | NR_034072.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009344.hg.1 | NR_034072.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009344.hg.1 | NR_034072.1 | Refseq |
