Assay Details
| Gene Symbol: SNRPE | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001304464.1 | NP_001291393.1 | 1 - 2 | 159 | 100 | |
| GenBank mRNA | AK307255.1 | - | 1 - 2 | 145 | 100 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001304464.1 transcripts.
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1600009653.hg.1 | ENST00000338402.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009653.hg.1 | ENST00000338402.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011276.hg.1 | ENST00000367208.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011276.hg.1 | ENST00000367208.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011276.hg.1 | ENST00000367208.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011276.hg.1 | ENST00000367208.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011276.hg.1 | ENST00000367208.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011276.hg.1 | ENST00000414487.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011276.hg.1 | ENST00000414487.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011276.hg.1 | ENST00000414487.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011276.hg.1 | ENST00000414487.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011276.hg.1 | ENST00000414487.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011276.hg.1 | ENST00000414487.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011276.hg.1 | ENST00000414487.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011276.hg.1 | ENST00000414487.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011276.hg.1 | ENST00000414487.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011276.hg.1 | ENST00000469451.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011276.hg.1 | ENST00000469451.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011276.hg.1 | ENST00000469451.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011276.hg.1 | ENST00000470492.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011276.hg.1 | ENST00000470492.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011276.hg.1 | ENST00000470492.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011276.hg.1 | ENST00000470492.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011276.hg.1 | ENST00000470492.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011276.hg.1 | ENST00000470492.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011276.hg.1 | ENST00000470492.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011276.hg.1 | ENST00000470492.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011276.hg.1 | ENST00000475035.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011276.hg.1 | ENST00000475035.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011276.hg.1 | ENST00000475035.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011276.hg.1 | ENST00000475035.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011276.hg.1 | ENST00000475035.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011276.hg.1 | ENST00000475035.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011276.hg.1 | ENST00000475035.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011276.hg.1 | ENST00000475035.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011276.hg.1 | ENST00000475035.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011276.hg.1 | ENST00000483099.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011276.hg.1 | ENST00000483099.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011276.hg.1 | ENST00000483099.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011276.hg.1 | ENST00000483099.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011276.hg.1 | ENST00000483099.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011276.hg.1 | ENST00000483099.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011276.hg.1 | ENST00000483099.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011276.hg.1 | ENST00000483099.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011276.hg.1 | ENST00000483099.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011276.hg.1 | ENST00000483099.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011276.hg.1 | ENST00000483099.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011276.hg.1 | ENST00000483099.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011276.hg.1 | NM_001304464.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011276.hg.1 | NM_001304464.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011276.hg.1 | NM_001304464.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011276.hg.1 | NM_001304464.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011276.hg.1 | NM_001304464.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011276.hg.1 | NM_001304464.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011276.hg.1 | NM_001304464.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011276.hg.1 | NM_001304464.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011276.hg.1 | NM_001304464.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011276.hg.1 | NM_001304464.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011276.hg.1 | NM_001304464.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011276.hg.1 | NM_003094.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011276.hg.1 | NM_003094.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011276.hg.1 | NM_003094.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011276.hg.1 | NM_003094.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011276.hg.1 | NM_003094.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011276.hg.1 | NM_003094.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011276.hg.1 | NM_003094.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011276.hg.1 | NM_003094.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011276.hg.1 | NM_003094.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011276.hg.1 | NM_003094.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011276.hg.1 | NR_130746.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011276.hg.1 | NR_130746.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011276.hg.1 | NR_130746.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011276.hg.1 | NR_130746.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011276.hg.1 | NR_130746.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011276.hg.1 | NR_130746.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011276.hg.1 | NR_130746.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011276.hg.1 | NR_130746.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011276.hg.1 | NR_130746.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011276.hg.1 | NR_130746.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011276.hg.1 | ENST00000367208.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011276.hg.1 | ENST00000367208.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011276.hg.1 | ENST00000367208.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011276.hg.1 | ENST00000367208.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011276.hg.1 | ENST00000367208.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011276.hg.1 | ENST00000414487.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011276.hg.1 | ENST00000414487.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011276.hg.1 | ENST00000414487.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011276.hg.1 | ENST00000414487.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011276.hg.1 | ENST00000414487.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011276.hg.1 | ENST00000414487.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011276.hg.1 | ENST00000414487.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011276.hg.1 | ENST00000414487.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011276.hg.1 | ENST00000414487.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011276.hg.1 | ENST00000469451.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011276.hg.1 | ENST00000469451.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011276.hg.1 | ENST00000469451.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011276.hg.1 | ENST00000470492.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011276.hg.1 | ENST00000470492.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011276.hg.1 | ENST00000470492.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011276.hg.1 | ENST00000470492.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011276.hg.1 | ENST00000470492.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011276.hg.1 | ENST00000470492.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011276.hg.1 | ENST00000470492.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011276.hg.1 | ENST00000470492.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011276.hg.1 | ENST00000475035.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011276.hg.1 | ENST00000475035.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011276.hg.1 | ENST00000475035.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011276.hg.1 | ENST00000475035.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011276.hg.1 | ENST00000475035.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011276.hg.1 | ENST00000475035.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011276.hg.1 | ENST00000475035.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011276.hg.1 | ENST00000475035.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011276.hg.1 | ENST00000475035.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011276.hg.1 | ENST00000483099.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011276.hg.1 | ENST00000483099.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011276.hg.1 | ENST00000483099.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011276.hg.1 | ENST00000483099.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011276.hg.1 | ENST00000483099.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011276.hg.1 | ENST00000483099.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011276.hg.1 | ENST00000483099.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011276.hg.1 | ENST00000483099.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011276.hg.1 | ENST00000483099.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011276.hg.1 | ENST00000483099.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011276.hg.1 | ENST00000483099.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011276.hg.1 | ENST00000483099.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011276.hg.1 | NM_001304464.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011276.hg.1 | NM_001304464.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011276.hg.1 | NM_001304464.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011276.hg.1 | NM_001304464.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011276.hg.1 | NM_001304464.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011276.hg.1 | NM_001304464.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011276.hg.1 | NM_001304464.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011276.hg.1 | NM_001304464.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011276.hg.1 | NM_001304464.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011276.hg.1 | NM_001304464.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011276.hg.1 | NM_001304464.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011276.hg.1 | NM_003094.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011276.hg.1 | NM_003094.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011276.hg.1 | NM_003094.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011276.hg.1 | NM_003094.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011276.hg.1 | NM_003094.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011276.hg.1 | NM_003094.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011276.hg.1 | NM_003094.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011276.hg.1 | NM_003094.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011276.hg.1 | NM_003094.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011276.hg.1 | NM_003094.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011276.hg.1 | NR_130746.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011276.hg.1 | NR_130746.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011276.hg.1 | NR_130746.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011276.hg.1 | NR_130746.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011276.hg.1 | NR_130746.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011276.hg.1 | NR_130746.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011276.hg.1 | NR_130746.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011276.hg.1 | NR_130746.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011276.hg.1 | NR_130746.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011276.hg.1 | NR_130746.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009653.hg.1 | ENST00000338402.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009653.hg.1 | ENST00000338402.6 | Ensembl |

SNRPE
Genomic Map