Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015596.hg.1 |
ENST00000604000.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015596.hg.1 |
ENST00000604000.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015596.hg.1 |
ENST00000604000.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015596.hg.1 |
ENST00000604000.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015596.hg.1 |
ENST00000604000.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015596.hg.1 |
ENST00000604000.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015596.hg.1 |
ENST00000604000.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015596.hg.1 |
ENST00000604000.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015596.hg.1 |
ENST00000604000.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015596.hg.1 |
ENST00000604000.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015596.hg.1 |
ENST00000604000.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015596.hg.1 |
ENST00000604000.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015596.hg.1 |
NM_153713.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015596.hg.1 |
NM_153713.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015596.hg.1 |
NM_153713.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015596.hg.1 |
NM_153713.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015596.hg.1 |
NM_153713.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015596.hg.1 |
NM_153713.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015596.hg.1 |
NM_153713.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015596.hg.1 |
NM_153713.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015596.hg.1 |
NM_153713.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015596.hg.1 |
NM_153713.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015596.hg.1 |
NM_153713.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015596.hg.1 |
NM_153713.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015596.hg.1 |
ENST00000604000.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015596.hg.1 |
ENST00000604000.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015596.hg.1 |
ENST00000604000.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015596.hg.1 |
ENST00000604000.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015596.hg.1 |
ENST00000604000.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015596.hg.1 |
ENST00000604000.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015596.hg.1 |
ENST00000604000.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015596.hg.1 |
ENST00000604000.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015596.hg.1 |
ENST00000604000.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015596.hg.1 |
ENST00000604000.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015596.hg.1 |
ENST00000604000.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015596.hg.1 |
ENST00000604000.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015596.hg.1 |
NM_153713.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015596.hg.1 |
NM_153713.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015596.hg.1 |
NM_153713.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015596.hg.1 |
NM_153713.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015596.hg.1 |
NM_153713.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015596.hg.1 |
NM_153713.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015596.hg.1 |
NM_153713.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015596.hg.1 |
NM_153713.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015596.hg.1 |
NM_153713.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015596.hg.1 |
NM_153713.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015596.hg.1 |
NM_153713.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015596.hg.1 |
NM_153713.2 |
Refseq |