Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010028.hg.1 |
ENST00000368742.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010028.hg.1 |
ENST00000368742.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010028.hg.1 |
ENST00000368742.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010028.hg.1 |
NM_000427.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010028.hg.1 |
NM_000427.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100010028.hg.1 |
NM_000427.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010028.hg.1 |
ENST00000368742.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010028.hg.1 |
ENST00000368742.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010028.hg.1 |
ENST00000368742.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100010028.hg.1 |
NM_000427.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010028.hg.1 |
NM_000427.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100010028.hg.1 |
NM_000427.2 |
Refseq |