Assay Details
| Gene Symbol: LOC283028 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_136644.1 | 1 | 3779 | 147 | ||
| GenBank mRNA | AF146695.1 | - | 1 | 721 | 147 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC283028| Gene Symbol: LOC283028 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_136644.1 | 1 | 3779 | 147 | ||
| GenBank mRNA | AF146695.1 | - | 1 | 721 | 147 | |
Set Membership: |
|