Assay Details
| Gene Symbol: SPATA21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_011541407.2 | XP_011539709.1 | 1 - 2 | 269 | 91 | |
| XM_011541417.2 | XP_011539719.1 | 1 - 2 | 269 | 91 | ||
| XM_017001216.1 | XP_016856705.1 | 1 - 2 | 269 | 91 | ||
| XM_017001218.1 | XP_016856707.1 | 1 - 2 | 269 | 91 | ||
| XM_017001219.1 | XP_016856708.1 | 1 - 2 | 269 | 91 | ||
| GenBank mRNA | AK128106.1 | - | 1 - 2 | 269 | 91 | |
| BC022039.1 | - | 1 - 2 | 297 | 91 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK128106.1, BC022039.1, XM_011541407.2, XM_011541417.2, XM_017001216.1, XM_017001218.1, XM_017001219.1 transcripts.
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0100013042.hg.1 | ENST00000335496.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013042.hg.1 | ENST00000335496.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013042.hg.1 | ENST00000335496.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013042.hg.1 | ENST00000335496.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013042.hg.1 | ENST00000335496.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013042.hg.1 | ENST00000335496.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013042.hg.1 | ENST00000335496.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013042.hg.1 | ENST00000335496.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013042.hg.1 | ENST00000335496.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013042.hg.1 | ENST00000335496.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013042.hg.1 | ENST00000335496.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013042.hg.1 | ENST00000335496.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013042.hg.1 | ENST00000335496.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013042.hg.1 | ENST00000335496.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013042.hg.1 | ENST00000335496.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013042.hg.1 | ENST00000335496.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013042.hg.1 | ENST00000375577.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013042.hg.1 | ENST00000375577.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013042.hg.1 | ENST00000375577.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013042.hg.1 | ENST00000375577.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013042.hg.1 | ENST00000375577.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013042.hg.1 | ENST00000375577.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013042.hg.1 | ENST00000375577.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013042.hg.1 | ENST00000375577.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013042.hg.1 | ENST00000375577.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013042.hg.1 | ENST00000466212.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013042.hg.1 | ENST00000466212.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013042.hg.1 | ENST00000466212.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013042.hg.1 | ENST00000466212.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013042.hg.1 | ENST00000466212.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013042.hg.1 | ENST00000466212.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013042.hg.1 | ENST00000466212.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013042.hg.1 | ENST00000466212.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013042.hg.1 | ENST00000466212.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013042.hg.1 | ENST00000466212.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013042.hg.1 | ENST00000466212.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013042.hg.1 | ENST00000466212.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013042.hg.1 | ENST00000466212.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013042.hg.1 | ENST00000466212.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013042.hg.1 | ENST00000466212.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013042.hg.1 | ENST00000466212.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013042.hg.1 | ENST00000466212.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013042.hg.1 | ENST00000466212.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013042.hg.1 | ENST00000466212.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013042.hg.1 | ENST00000466212.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013042.hg.1 | ENST00000466212.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013042.hg.1 | ENST00000491418.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013042.hg.1 | ENST00000491418.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013042.hg.1 | ENST00000491418.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013042.hg.1 | ENST00000491418.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013042.hg.1 | ENST00000540400.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013042.hg.1 | ENST00000540400.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013042.hg.1 | ENST00000540400.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013042.hg.1 | ENST00000540400.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013042.hg.1 | ENST00000540400.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013042.hg.1 | ENST00000540400.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013042.hg.1 | ENST00000540400.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013042.hg.1 | ENST00000540400.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013042.hg.1 | ENST00000540400.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013042.hg.1 | ENST00000540400.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013042.hg.1 | ENST00000540400.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013042.hg.1 | ENST00000540400.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013042.hg.1 | ENST00000612240.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013042.hg.1 | ENST00000612240.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013042.hg.1 | ENST00000612240.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013042.hg.1 | ENST00000612240.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013042.hg.1 | ENST00000612240.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013042.hg.1 | NM_198546.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013042.hg.1 | NM_198546.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013042.hg.1 | NM_198546.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013042.hg.1 | NM_198546.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013042.hg.1 | NM_198546.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013042.hg.1 | NM_198546.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013042.hg.1 | NM_198546.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013042.hg.1 | NM_198546.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013042.hg.1 | NM_198546.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013042.hg.1 | NM_198546.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013042.hg.1 | NM_198546.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013042.hg.1 | NM_198546.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013042.hg.1 | NM_198546.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013042.hg.1 | NM_198546.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013042.hg.1 | NM_198546.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013042.hg.1 | NM_198546.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013042.hg.1 | ENST00000335496.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013042.hg.1 | ENST00000335496.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013042.hg.1 | ENST00000335496.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013042.hg.1 | ENST00000335496.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013042.hg.1 | ENST00000335496.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013042.hg.1 | ENST00000335496.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013042.hg.1 | ENST00000335496.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013042.hg.1 | ENST00000335496.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013042.hg.1 | ENST00000335496.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013042.hg.1 | ENST00000335496.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013042.hg.1 | ENST00000335496.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013042.hg.1 | ENST00000335496.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013042.hg.1 | ENST00000335496.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013042.hg.1 | ENST00000335496.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013042.hg.1 | ENST00000335496.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013042.hg.1 | ENST00000335496.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013042.hg.1 | ENST00000375577.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013042.hg.1 | ENST00000375577.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013042.hg.1 | ENST00000375577.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013042.hg.1 | ENST00000375577.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013042.hg.1 | ENST00000375577.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013042.hg.1 | ENST00000375577.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013042.hg.1 | ENST00000375577.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013042.hg.1 | ENST00000375577.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013042.hg.1 | ENST00000375577.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013042.hg.1 | ENST00000466212.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013042.hg.1 | ENST00000466212.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013042.hg.1 | ENST00000466212.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013042.hg.1 | ENST00000466212.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013042.hg.1 | ENST00000466212.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013042.hg.1 | ENST00000466212.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013042.hg.1 | ENST00000466212.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013042.hg.1 | ENST00000466212.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013042.hg.1 | ENST00000466212.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013042.hg.1 | ENST00000466212.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013042.hg.1 | ENST00000466212.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013042.hg.1 | ENST00000466212.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013042.hg.1 | ENST00000466212.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013042.hg.1 | ENST00000466212.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013042.hg.1 | ENST00000466212.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013042.hg.1 | ENST00000466212.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013042.hg.1 | ENST00000466212.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013042.hg.1 | ENST00000466212.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013042.hg.1 | ENST00000466212.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013042.hg.1 | ENST00000466212.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013042.hg.1 | ENST00000466212.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013042.hg.1 | ENST00000491418.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013042.hg.1 | ENST00000491418.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013042.hg.1 | ENST00000491418.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013042.hg.1 | ENST00000491418.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013042.hg.1 | ENST00000540400.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013042.hg.1 | ENST00000540400.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013042.hg.1 | ENST00000540400.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013042.hg.1 | ENST00000540400.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013042.hg.1 | ENST00000540400.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013042.hg.1 | ENST00000540400.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013042.hg.1 | ENST00000540400.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013042.hg.1 | ENST00000540400.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013042.hg.1 | ENST00000540400.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013042.hg.1 | ENST00000540400.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013042.hg.1 | ENST00000540400.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013042.hg.1 | ENST00000540400.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013042.hg.1 | ENST00000612240.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013042.hg.1 | ENST00000612240.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013042.hg.1 | ENST00000612240.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013042.hg.1 | ENST00000612240.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013042.hg.1 | ENST00000612240.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013042.hg.1 | NM_198546.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013042.hg.1 | NM_198546.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013042.hg.1 | NM_198546.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013042.hg.1 | NM_198546.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013042.hg.1 | NM_198546.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013042.hg.1 | NM_198546.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013042.hg.1 | NM_198546.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013042.hg.1 | NM_198546.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013042.hg.1 | NM_198546.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013042.hg.1 | NM_198546.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013042.hg.1 | NM_198546.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013042.hg.1 | NM_198546.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013042.hg.1 | NM_198546.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013042.hg.1 | NM_198546.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013042.hg.1 | NM_198546.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013042.hg.1 | NM_198546.1 | Refseq |

SPATA21
Genomic Map