Assay Details
| Gene Symbol: SPATA21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC022039.1 | - | 4 - 5 | 830 | 111 | |
More Information
Set Membership: |
|
Panther Classification:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0100013042.hg.1 | ENST00000335496.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013042.hg.1 | ENST00000335496.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013042.hg.1 | ENST00000335496.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013042.hg.1 | ENST00000335496.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013042.hg.1 | ENST00000335496.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013042.hg.1 | ENST00000335496.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013042.hg.1 | ENST00000335496.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013042.hg.1 | ENST00000335496.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013042.hg.1 | ENST00000335496.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013042.hg.1 | ENST00000335496.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013042.hg.1 | ENST00000335496.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013042.hg.1 | ENST00000335496.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013042.hg.1 | ENST00000335496.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013042.hg.1 | ENST00000335496.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013042.hg.1 | ENST00000335496.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013042.hg.1 | ENST00000335496.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013042.hg.1 | ENST00000375577.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013042.hg.1 | ENST00000375577.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013042.hg.1 | ENST00000375577.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013042.hg.1 | ENST00000375577.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013042.hg.1 | ENST00000375577.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013042.hg.1 | ENST00000375577.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013042.hg.1 | ENST00000375577.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013042.hg.1 | ENST00000375577.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013042.hg.1 | ENST00000375577.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013042.hg.1 | ENST00000466212.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013042.hg.1 | ENST00000466212.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013042.hg.1 | ENST00000466212.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013042.hg.1 | ENST00000466212.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013042.hg.1 | ENST00000466212.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013042.hg.1 | ENST00000466212.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013042.hg.1 | ENST00000466212.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013042.hg.1 | ENST00000466212.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013042.hg.1 | ENST00000466212.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013042.hg.1 | ENST00000466212.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013042.hg.1 | ENST00000466212.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013042.hg.1 | ENST00000466212.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013042.hg.1 | ENST00000466212.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013042.hg.1 | ENST00000466212.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013042.hg.1 | ENST00000466212.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013042.hg.1 | ENST00000466212.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013042.hg.1 | ENST00000466212.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013042.hg.1 | ENST00000466212.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013042.hg.1 | ENST00000466212.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013042.hg.1 | ENST00000466212.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013042.hg.1 | ENST00000466212.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013042.hg.1 | ENST00000491418.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013042.hg.1 | ENST00000491418.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013042.hg.1 | ENST00000491418.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013042.hg.1 | ENST00000491418.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013042.hg.1 | ENST00000540400.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013042.hg.1 | ENST00000540400.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013042.hg.1 | ENST00000540400.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013042.hg.1 | ENST00000540400.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013042.hg.1 | ENST00000540400.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013042.hg.1 | ENST00000540400.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013042.hg.1 | ENST00000540400.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013042.hg.1 | ENST00000540400.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013042.hg.1 | ENST00000540400.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013042.hg.1 | ENST00000540400.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013042.hg.1 | ENST00000540400.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013042.hg.1 | ENST00000540400.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013042.hg.1 | ENST00000612240.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013042.hg.1 | ENST00000612240.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013042.hg.1 | ENST00000612240.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013042.hg.1 | ENST00000612240.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013042.hg.1 | ENST00000612240.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013042.hg.1 | NM_198546.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013042.hg.1 | NM_198546.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013042.hg.1 | NM_198546.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013042.hg.1 | NM_198546.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013042.hg.1 | NM_198546.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013042.hg.1 | NM_198546.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013042.hg.1 | NM_198546.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013042.hg.1 | NM_198546.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013042.hg.1 | NM_198546.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013042.hg.1 | NM_198546.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013042.hg.1 | NM_198546.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013042.hg.1 | NM_198546.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013042.hg.1 | NM_198546.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013042.hg.1 | NM_198546.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013042.hg.1 | NM_198546.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100013042.hg.1 | NM_198546.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013042.hg.1 | ENST00000335496.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013042.hg.1 | ENST00000335496.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013042.hg.1 | ENST00000335496.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013042.hg.1 | ENST00000335496.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013042.hg.1 | ENST00000335496.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013042.hg.1 | ENST00000335496.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013042.hg.1 | ENST00000335496.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013042.hg.1 | ENST00000335496.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013042.hg.1 | ENST00000335496.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013042.hg.1 | ENST00000335496.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013042.hg.1 | ENST00000335496.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013042.hg.1 | ENST00000335496.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013042.hg.1 | ENST00000335496.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013042.hg.1 | ENST00000335496.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013042.hg.1 | ENST00000335496.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013042.hg.1 | ENST00000335496.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013042.hg.1 | ENST00000375577.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013042.hg.1 | ENST00000375577.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013042.hg.1 | ENST00000375577.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013042.hg.1 | ENST00000375577.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013042.hg.1 | ENST00000375577.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013042.hg.1 | ENST00000375577.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013042.hg.1 | ENST00000375577.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013042.hg.1 | ENST00000375577.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013042.hg.1 | ENST00000375577.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013042.hg.1 | ENST00000466212.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013042.hg.1 | ENST00000466212.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013042.hg.1 | ENST00000466212.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013042.hg.1 | ENST00000466212.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013042.hg.1 | ENST00000466212.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013042.hg.1 | ENST00000466212.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013042.hg.1 | ENST00000466212.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013042.hg.1 | ENST00000466212.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013042.hg.1 | ENST00000466212.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013042.hg.1 | ENST00000466212.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013042.hg.1 | ENST00000466212.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013042.hg.1 | ENST00000466212.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013042.hg.1 | ENST00000466212.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013042.hg.1 | ENST00000466212.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013042.hg.1 | ENST00000466212.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013042.hg.1 | ENST00000466212.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013042.hg.1 | ENST00000466212.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013042.hg.1 | ENST00000466212.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013042.hg.1 | ENST00000466212.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013042.hg.1 | ENST00000466212.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013042.hg.1 | ENST00000466212.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013042.hg.1 | ENST00000491418.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013042.hg.1 | ENST00000491418.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013042.hg.1 | ENST00000491418.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013042.hg.1 | ENST00000491418.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013042.hg.1 | ENST00000540400.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013042.hg.1 | ENST00000540400.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013042.hg.1 | ENST00000540400.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013042.hg.1 | ENST00000540400.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013042.hg.1 | ENST00000540400.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013042.hg.1 | ENST00000540400.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013042.hg.1 | ENST00000540400.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013042.hg.1 | ENST00000540400.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013042.hg.1 | ENST00000540400.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013042.hg.1 | ENST00000540400.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013042.hg.1 | ENST00000540400.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013042.hg.1 | ENST00000540400.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013042.hg.1 | ENST00000612240.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013042.hg.1 | ENST00000612240.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013042.hg.1 | ENST00000612240.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013042.hg.1 | ENST00000612240.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013042.hg.1 | ENST00000612240.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100013042.hg.1 | NM_198546.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013042.hg.1 | NM_198546.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013042.hg.1 | NM_198546.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013042.hg.1 | NM_198546.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013042.hg.1 | NM_198546.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013042.hg.1 | NM_198546.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013042.hg.1 | NM_198546.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013042.hg.1 | NM_198546.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013042.hg.1 | NM_198546.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013042.hg.1 | NM_198546.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013042.hg.1 | NM_198546.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013042.hg.1 | NM_198546.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013042.hg.1 | NM_198546.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013042.hg.1 | NM_198546.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013042.hg.1 | NM_198546.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100013042.hg.1 | NM_198546.1 | Refseq |

SPATA21
Genomic Map