Assay Details
| Gene Symbol: MEGF6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_011540885.1 | XP_011539187.1 | 1 - 2 | 452 | 93 | |
| XM_011540886.1 | XP_011539188.1 | 1 - 2 | 452 | 93 | ||
| XM_011540888.2 | XP_011539190.1 | 1 - 2 | 452 | 93 | ||
| XM_017000533.1 | XP_016856022.1 | 1 - 2 | 452 | 93 | ||
More Information
Set Membership: |
|

MEGF6
Genomic Map