Assay Details
Gene Symbol: SMIM12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_011540554.2 | XP_011538856.2 | 1 - 2 | 1340 | 91 | |
GenBank mRNA | BC034598.1 | - | 1 - 2 | 1885 | 91 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC034598.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0100018430.hg.1 | ENST00000417239.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018430.hg.1 | ENST00000417239.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018430.hg.1 | ENST00000417239.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018430.hg.1 | ENST00000417239.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018430.hg.1 | ENST00000423898.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018430.hg.1 | ENST00000423898.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018430.hg.1 | ENST00000423898.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018430.hg.1 | ENST00000423898.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018430.hg.1 | ENST00000423898.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018430.hg.1 | ENST00000423898.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018430.hg.1 | ENST00000446026.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018430.hg.1 | ENST00000446026.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018430.hg.1 | ENST00000446026.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018430.hg.1 | ENST00000446026.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018430.hg.1 | ENST00000446026.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018430.hg.1 | ENST00000446026.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018430.hg.1 | ENST00000446026.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018430.hg.1 | ENST00000446026.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018430.hg.1 | ENST00000456842.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018430.hg.1 | ENST00000456842.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018430.hg.1 | ENST00000456842.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018430.hg.1 | ENST00000456842.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018430.hg.1 | ENST00000456842.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018430.hg.1 | ENST00000456842.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018430.hg.1 | ENST00000521580.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018430.hg.1 | ENST00000521580.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018430.hg.1 | ENST00000521580.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018430.hg.1 | ENST00000521580.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018430.hg.1 | ENST00000521580.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018430.hg.1 | ENST00000521580.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018430.hg.1 | ENST00000521580.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018430.hg.1 | ENST00000521580.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018430.hg.1 | ENST00000521580.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018430.hg.1 | NM_001164824.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018430.hg.1 | NM_001164824.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018430.hg.1 | NM_001164824.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018430.hg.1 | NM_001164824.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018430.hg.1 | NM_001164824.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018430.hg.1 | NM_001164824.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018430.hg.1 | NM_001164824.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018430.hg.1 | NM_001164824.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018430.hg.1 | NM_001164824.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018430.hg.1 | NM_001164824.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018430.hg.1 | NM_001164825.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018430.hg.1 | NM_001164825.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018430.hg.1 | NM_001164825.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018430.hg.1 | NM_001164825.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018430.hg.1 | NM_001164825.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018430.hg.1 | NM_001164825.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018430.hg.1 | NM_001164825.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018430.hg.1 | NM_001164825.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018430.hg.1 | NM_001164825.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018430.hg.1 | NM_001164825.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018430.hg.1 | NM_001164825.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018430.hg.1 | NM_138428.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018430.hg.1 | NM_138428.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018430.hg.1 | NM_138428.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018430.hg.1 | NM_138428.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018430.hg.1 | NM_138428.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018430.hg.1 | NM_138428.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018430.hg.1 | NM_138428.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018430.hg.1 | NM_138428.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018430.hg.1 | NM_138428.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018430.hg.1 | NM_138428.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018430.hg.1 | ENST00000417239.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018430.hg.1 | ENST00000417239.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018430.hg.1 | ENST00000417239.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018430.hg.1 | ENST00000417239.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018430.hg.1 | ENST00000423898.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018430.hg.1 | ENST00000423898.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018430.hg.1 | ENST00000423898.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018430.hg.1 | ENST00000423898.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018430.hg.1 | ENST00000423898.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018430.hg.1 | ENST00000423898.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018430.hg.1 | ENST00000446026.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018430.hg.1 | ENST00000446026.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018430.hg.1 | ENST00000446026.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018430.hg.1 | ENST00000446026.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018430.hg.1 | ENST00000446026.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018430.hg.1 | ENST00000446026.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018430.hg.1 | ENST00000446026.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018430.hg.1 | ENST00000446026.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018430.hg.1 | ENST00000456842.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018430.hg.1 | ENST00000456842.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018430.hg.1 | ENST00000456842.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018430.hg.1 | ENST00000456842.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018430.hg.1 | ENST00000456842.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018430.hg.1 | ENST00000456842.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018430.hg.1 | ENST00000521580.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018430.hg.1 | ENST00000521580.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018430.hg.1 | ENST00000521580.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018430.hg.1 | ENST00000521580.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018430.hg.1 | ENST00000521580.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018430.hg.1 | ENST00000521580.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018430.hg.1 | ENST00000521580.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018430.hg.1 | ENST00000521580.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018430.hg.1 | ENST00000521580.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018430.hg.1 | NM_001164824.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018430.hg.1 | NM_001164824.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018430.hg.1 | NM_001164824.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018430.hg.1 | NM_001164824.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018430.hg.1 | NM_001164824.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018430.hg.1 | NM_001164824.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018430.hg.1 | NM_001164824.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018430.hg.1 | NM_001164824.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018430.hg.1 | NM_001164824.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018430.hg.1 | NM_001164824.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018430.hg.1 | NM_001164825.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018430.hg.1 | NM_001164825.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018430.hg.1 | NM_001164825.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018430.hg.1 | NM_001164825.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018430.hg.1 | NM_001164825.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018430.hg.1 | NM_001164825.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018430.hg.1 | NM_001164825.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018430.hg.1 | NM_001164825.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018430.hg.1 | NM_001164825.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018430.hg.1 | NM_001164825.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018430.hg.1 | NM_001164825.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018430.hg.1 | NM_138428.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018430.hg.1 | NM_138428.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018430.hg.1 | NM_138428.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018430.hg.1 | NM_138428.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018430.hg.1 | NM_138428.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018430.hg.1 | NM_138428.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018430.hg.1 | NM_138428.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018430.hg.1 | NM_138428.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018430.hg.1 | NM_138428.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018430.hg.1 | NM_138428.5 | Refseq |
