| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016373.hg.1 |
ENST00000440404.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016373.hg.1 |
ENST00000440404.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016373.hg.1 |
ENST00000440404.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016373.hg.1 |
ENST00000440404.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016373.hg.1 |
ENST00000440404.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016373.hg.1 |
ENST00000440404.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016373.hg.1 |
ENST00000440404.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016373.hg.1 |
ENST00000440404.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016373.hg.1 |
ENST00000440404.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016373.hg.1 |
ENST00000440404.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016373.hg.1 |
ENST00000451439.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016373.hg.1 |
ENST00000451439.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016373.hg.1 |
ENST00000451439.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016373.hg.1 |
NR_026957.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016373.hg.1 |
NR_026957.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016373.hg.1 |
NR_026957.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016373.hg.1 |
NR_026957.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016373.hg.1 |
NR_026957.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016373.hg.1 |
NR_026957.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100016373.hg.1 |
NR_026957.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100016373.hg.1 |
ENST00000440404.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016373.hg.1 |
ENST00000440404.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016373.hg.1 |
ENST00000440404.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016373.hg.1 |
ENST00000440404.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016373.hg.1 |
ENST00000440404.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016373.hg.1 |
ENST00000440404.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016373.hg.1 |
ENST00000440404.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016373.hg.1 |
ENST00000440404.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016373.hg.1 |
ENST00000440404.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016373.hg.1 |
ENST00000440404.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016373.hg.1 |
ENST00000451439.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016373.hg.1 |
ENST00000451439.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016373.hg.1 |
ENST00000451439.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100016373.hg.1 |
NR_026957.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100016373.hg.1 |
NR_026957.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100016373.hg.1 |
NR_026957.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100016373.hg.1 |
NR_026957.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100016373.hg.1 |
NR_026957.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100016373.hg.1 |
NR_026957.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100016373.hg.1 |
NR_026957.1 |
Refseq |