Assay Details
| Gene Symbol: SPATA17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005273052.1 | XP_005273109.1 | 10 - 11 | 1126 | 143 | |
| XM_006711165.3 | XP_006711228.1 | 9 - 10 | 1084 | 143 | ||
More Information
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0100011611.hg.1 | ENST00000366933.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011611.hg.1 | ENST00000366933.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011611.hg.1 | ENST00000366933.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011611.hg.1 | ENST00000366933.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011611.hg.1 | ENST00000366933.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011611.hg.1 | ENST00000366933.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011611.hg.1 | ENST00000366933.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011611.hg.1 | ENST00000366933.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011611.hg.1 | ENST00000366933.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011611.hg.1 | ENST00000366933.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011611.hg.1 | ENST00000366933.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011611.hg.1 | ENST00000366933.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011611.hg.1 | ENST00000366933.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011611.hg.1 | ENST00000366933.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011611.hg.1 | ENST00000366933.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011611.hg.1 | ENST00000366933.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011611.hg.1 | ENST00000366933.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011611.hg.1 | ENST00000366933.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011611.hg.1 | ENST00000470448.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011611.hg.1 | ENST00000470448.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011611.hg.1 | ENST00000470448.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011611.hg.1 | ENST00000470448.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011611.hg.1 | ENST00000470448.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011611.hg.1 | ENST00000470448.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011611.hg.1 | ENST00000470448.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011611.hg.1 | ENST00000470448.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011611.hg.1 | ENST00000470448.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011611.hg.1 | ENST00000470448.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011611.hg.1 | ENST00000471021.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011611.hg.1 | ENST00000471021.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011611.hg.1 | ENST00000471021.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011611.hg.1 | ENST00000471021.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011611.hg.1 | ENST00000471021.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011611.hg.1 | ENST00000471021.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011611.hg.1 | ENST00000491809.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011611.hg.1 | ENST00000491809.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011611.hg.1 | ENST00000491809.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011611.hg.1 | ENST00000492747.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011611.hg.1 | ENST00000492747.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011611.hg.1 | ENST00000492747.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011611.hg.1 | ENST00000492747.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011611.hg.1 | ENST00000492747.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011611.hg.1 | ENST00000492747.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011611.hg.1 | ENST00000492747.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011611.hg.1 | NM_138796.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011611.hg.1 | NM_138796.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011611.hg.1 | NM_138796.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011611.hg.1 | NM_138796.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011611.hg.1 | NM_138796.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011611.hg.1 | NM_138796.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011611.hg.1 | NM_138796.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011611.hg.1 | NM_138796.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011611.hg.1 | NM_138796.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011611.hg.1 | NM_138796.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011611.hg.1 | NM_138796.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011611.hg.1 | NM_138796.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011611.hg.1 | NM_138796.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011611.hg.1 | NM_138796.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011611.hg.1 | NM_138796.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011611.hg.1 | NM_138796.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011611.hg.1 | NM_138796.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011611.hg.1 | ENST00000366933.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011611.hg.1 | ENST00000366933.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011611.hg.1 | ENST00000366933.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011611.hg.1 | ENST00000366933.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011611.hg.1 | ENST00000366933.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011611.hg.1 | ENST00000366933.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011611.hg.1 | ENST00000366933.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011611.hg.1 | ENST00000366933.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011611.hg.1 | ENST00000366933.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011611.hg.1 | ENST00000366933.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011611.hg.1 | ENST00000366933.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011611.hg.1 | ENST00000366933.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011611.hg.1 | ENST00000366933.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011611.hg.1 | ENST00000366933.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011611.hg.1 | ENST00000366933.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011611.hg.1 | ENST00000366933.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011611.hg.1 | ENST00000366933.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011611.hg.1 | ENST00000366933.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011611.hg.1 | ENST00000470448.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011611.hg.1 | ENST00000470448.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011611.hg.1 | ENST00000470448.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011611.hg.1 | ENST00000470448.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011611.hg.1 | ENST00000470448.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011611.hg.1 | ENST00000470448.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011611.hg.1 | ENST00000470448.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011611.hg.1 | ENST00000470448.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011611.hg.1 | ENST00000470448.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011611.hg.1 | ENST00000470448.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011611.hg.1 | ENST00000471021.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011611.hg.1 | ENST00000471021.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011611.hg.1 | ENST00000471021.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011611.hg.1 | ENST00000471021.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011611.hg.1 | ENST00000471021.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011611.hg.1 | ENST00000471021.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011611.hg.1 | ENST00000491809.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011611.hg.1 | ENST00000491809.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011611.hg.1 | ENST00000491809.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011611.hg.1 | ENST00000492747.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011611.hg.1 | ENST00000492747.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011611.hg.1 | ENST00000492747.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011611.hg.1 | ENST00000492747.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011611.hg.1 | ENST00000492747.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011611.hg.1 | ENST00000492747.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011611.hg.1 | ENST00000492747.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011611.hg.1 | NM_138796.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011611.hg.1 | NM_138796.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011611.hg.1 | NM_138796.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011611.hg.1 | NM_138796.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011611.hg.1 | NM_138796.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011611.hg.1 | NM_138796.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011611.hg.1 | NM_138796.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011611.hg.1 | NM_138796.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011611.hg.1 | NM_138796.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011611.hg.1 | NM_138796.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011611.hg.1 | NM_138796.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011611.hg.1 | NM_138796.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011611.hg.1 | NM_138796.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011611.hg.1 | NM_138796.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011611.hg.1 | NM_138796.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011611.hg.1 | NM_138796.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011611.hg.1 | NM_138796.3 | Refseq |

SPATA17
Genomic Map