Assay Details
| Gene Symbol: RHCE | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_011541888.2 | XP_011540190.1 | 2 - 3 | 739 | 109 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_011541888.2 transcripts.
Set Membership: |
|

RHCE
Genomic Map