Assay Details
| Gene Symbol: GNL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001323623.1 | NP_001310552.1 | 7 - 8 | 959 | 112 | |
| NM_001323624.1 | NP_001310553.1 | 6 - 7 | 774 | 112 | ||
| NM_013285.2 | NP_037417.1 | 7 - 8 | 959 | 112 | ||
| GenBank mRNA | AK298358.1 | - | 7 - 8 | 869 | 112 | |
| AK312915.1 | - | 7 - 8 | 913 | 112 | ||
| BC000107.2 | - | 7 - 8 | 895 | 3506131 | 112 | |
| BC009250.2 | - | 7 - 8 | 895 | 2960125 | 112 | |
| L05425.1 | - | 7 - 8 | 875 | 112 | ||
More Information
Set Membership: |
|

GNL2
Genomic Map