Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018436.hg.1 |
ENST00000466927.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018436.hg.1 |
ENST00000466927.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018436.hg.1 |
ENST00000466927.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018436.hg.1 |
ENST00000474566.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018436.hg.1 |
ENST00000474566.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018436.hg.1 |
ENST00000474566.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018436.hg.1 |
ENST00000474566.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018436.hg.1 |
ENST00000474566.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018436.hg.1 |
ENST00000466927.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018436.hg.1 |
ENST00000466927.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018436.hg.1 |
ENST00000466927.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018436.hg.1 |
ENST00000474566.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018436.hg.1 |
ENST00000474566.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018436.hg.1 |
ENST00000474566.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018436.hg.1 |
ENST00000474566.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018436.hg.1 |
ENST00000474566.1 |
Ensembl |